Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32493042-32493315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO7
Entrez Gene ID
25793
Gene Name
F-box protein 7
Gene Aliases
FBX, FBX07, FBX7, PARK15, PKPS
UniGene
Hs.5912
Gene Chromosome Location
Chr.22: 32474682-32498831 on Build GRCh38
Transcripts
NM_001033024.1 NM_001257990.1 NM_012179.3 XM_011530106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1682231 c.901T>G p.F301V Chr.22: 32493275-32493275 on Build GRCh38
1751841 c.999C>T p.V333V Chr.22: 32493136-32493136 on Build GRCh38
3992119 c.1007C>A p.P336Q Chr.22: 32493144-32493144 on Build GRCh38
6876855 c.1032C>T p.F344F Chr.22: 32493169-32493169 on Build GRCh38
3992121 c.770C>A p.P257Q Chr.22: 32493144-32493144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780834791 Chr.22: 32493185-32493185 on Build GRCh38
rs754811367 Chr.22: 32493146-32493146 on Build GRCh38
rs764006085 Chr.22: 32493237-32493237 on Build GRCh38
rs750505885 Chr.22: 32493116-32493116 on Build GRCh38
rs778534727 Chr.22: 32493152-32493152 on Build GRCh38
rs949918227 Chr.22: 32493222-32493222 on Build GRCh38
rs554745494 Chr.22: 32493109-32493109 on Build GRCh38
rs779372239 Chr.22: 32493279-32493279 on Build GRCh38
rs111604607 Chr.22: 32493216-32493216 on Build GRCh38
rs964834640 Chr.22: 32493183-32493183 on Build GRCh38
rs759631116 Chr.22: 32493218-32493218 on Build GRCh38
rs375622637 Chr.22: 32493069-32493069 on Build GRCh38
rs754058938 Chr.22: 32493137-32493137 on Build GRCh38
rs762037477 Chr.22: 32493191-32493191 on Build GRCh38
rs770629287 Chr.22: 32493171-32493171 on Build GRCh38
rs767654983 Chr.22: 32493108-32493108 on Build GRCh38
rs78099169 Chr.22: 32493278-32493278 on Build GRCh38
rs563829719 Chr.22: 32493236-32493236 on Build GRCh38
rs760820410 Chr.22: 32493213-32493213 on Build GRCh38
rs745445263 Chr.22: 32493188-32493188 on Build GRCh38
rs774729953 Chr.22: 32493096-32493096 on Build GRCh38
rs73884930 Chr.22: 32493078-32493078 on Build GRCh38
rs151103559 Chr.22: 32493170-32493170 on Build GRCh38
rs199696887 Chr.22: 32493090-32493090 on Build GRCh38
rs77327421 Chr.22: 32493091-32493091 on Build GRCh38
rs758247538 Chr.22: 32493163-32493163 on Build GRCh38
rs761014790 Chr.22: 32493124-32493124 on Build GRCh38
rs751440649 Chr.22: 32493255-32493255 on Build GRCh38
rs769094344 Chr.22: 32493189-32493189 on Build GRCh38
rs766700888 Chr.22: 32493130-32493130 on Build GRCh38
rs771646258 Chr.22: 32493073-32493073 on Build GRCh38
rs752706445 Chr.22: 32493155-32493155 on Build GRCh38
rs768845416 Chr.22: 32493097-32493097 on Build GRCh38
rs375751043 Chr.22: 32493270-32493270 on Build GRCh38
rs774488219 Chr.22: 32493153-32493153 on Build GRCh38
rs71799110 Chr.22: 32493269-32493269 on Build GRCh38
rs371871260 Chr.22: 32493231-32493231 on Build GRCh38
rs774925692 Chr.22: 32493104-32493104 on Build GRCh38
rs376900914 Chr.22: 32493075-32493075 on Build GRCh38
rs777219226 Chr.22: 32493076-32493076 on Build GRCh38
rs758087537 Chr.22: 32493201-32493204 on Build GRCh38
rs730880272 Chr.22: 32493282-32493282 on Build GRCh38
rs925636794 Chr.22: 32493099-32493099 on Build GRCh38
rs201841499 Chr.22: 32493204-32493204 on Build GRCh38
rs141068487 Chr.22: 32493207-32493207 on Build GRCh38
rs762185670 Chr.22: 32493107-32493107 on Build GRCh38
rs780638544 Chr.22: 32493263-32493263 on Build GRCh38
rs752442011 Chr.22: 32493090-32493090 on Build GRCh38
rs772673410 Chr.22: 32493205-32493205 on Build GRCh38
rs765132485 Chr.22: 32493219-32493219 on Build GRCh38
rs766504416 Chr.22: 32493215-32493215 on Build GRCh38
rs147911892 Chr.22: 32493262-32493262 on Build GRCh38

View Full Product Details