Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109105929-109106200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf194
Entrez Gene ID
127003
Gene Name
chromosome 1 open reading frame 194
Gene Aliases
-
UniGene
Hs.446962
Gene Chromosome Location
Chr.1: 109105951-109113877 on Build GRCh38
Transcripts
NM_001122961.1 XM_005270448.3 XM_006710342.3 XM_006710343.3 XM_011540647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615214 c.442C>T p.R148* Chr.1: 109106060-109106060 on Build GRCh38
4768628 c.422C>A p.A141D Chr.1: 109106080-109106080 on Build GRCh38
4981242 c.409C>T p.P137S Chr.1: 109106093-109106093 on Build GRCh38
3975837 c.468C>T p.S156S Chr.1: 109106034-109106034 on Build GRCh38
6615215 c.478C>T p.R160* Chr.1: 109106060-109106060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374027904 Chr.1: 109106051-109106051 on Build GRCh38
rs918600904 Chr.1: 109106023-109106023 on Build GRCh38
rs536121053 Chr.1: 109105980-109105980 on Build GRCh38
rs768819069 Chr.1: 109106104-109106104 on Build GRCh38
rs919803895 Chr.1: 109106141-109106141 on Build GRCh38
rs754196688 Chr.1: 109106018-109106018 on Build GRCh38
rs565061535 Chr.1: 109106098-109106098 on Build GRCh38
rs370124504 Chr.1: 109106047-109106047 on Build GRCh38
rs751563016 Chr.1: 109106071-109106071 on Build GRCh38
rs17641032 Chr.1: 109106172-109106172 on Build GRCh38
rs550425594 Chr.1: 109106140-109106140 on Build GRCh38
rs779106469 Chr.1: 109106112-109106112 on Build GRCh38
rs766679425 Chr.1: 109106008-109106008 on Build GRCh38
rs761115597 Chr.1: 109106005-109106005 on Build GRCh38
rs572792451 Chr.1: 109106034-109106034 on Build GRCh38
rs150611714 Chr.1: 109105986-109105986 on Build GRCh38
rs768396395 Chr.1: 109106100-109106101 on Build GRCh38
rs1020384052 Chr.1: 109106038-109106038 on Build GRCh38
rs530853974 Chr.1: 109106135-109106135 on Build GRCh38
rs962726882 Chr.1: 109105992-109105992 on Build GRCh38
rs774344711 Chr.1: 109105985-109105985 on Build GRCh38
rs370368387 Chr.1: 109105987-109105987 on Build GRCh38
rs377663410 Chr.1: 109106059-109106059 on Build GRCh38
rs746984529 Chr.1: 109106091-109106092 on Build GRCh38
rs1043943277 Chr.1: 109106146-109106146 on Build GRCh38
rs1019442896 Chr.1: 109106085-109106085 on Build GRCh38
rs773718206 Chr.1: 109106002-109106002 on Build GRCh38
rs780981951 Chr.1: 109106132-109106132 on Build GRCh38
rs916725173 Chr.1: 109106072-109106072 on Build GRCh38
rs373641326 Chr.1: 109106129-109106129 on Build GRCh38
rs772156057 Chr.1: 109106126-109106126 on Build GRCh38
rs369665369 Chr.1: 109106074-109106074 on Build GRCh38
rs763717608 Chr.1: 109106043-109106043 on Build GRCh38
rs777360003 Chr.1: 109106060-109106060 on Build GRCh38
rs759918072 Chr.1: 109105973-109105973 on Build GRCh38
rs764925629 Chr.1: 109106028-109106028 on Build GRCh38
rs948247919 Chr.1: 109106113-109106113 on Build GRCh38
rs748307305 Chr.1: 109106117-109106117 on Build GRCh38
rs759158389 Chr.1: 109106021-109106021 on Build GRCh38
rs780890959 Chr.1: 109106094-109106094 on Build GRCh38
rs375722101 Chr.1: 109106044-109106044 on Build GRCh38

View Full Product Details