Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 23762065-23762314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL2
Entrez Gene ID
1993
Gene Name
ELAV like neuron-specific RNA binding protein 2
Gene Aliases
HEL-N1, HELN1, HUB
UniGene
Hs.166109
Gene Chromosome Location
Chr.9: 23690099-23850757 on Build GRCh38
Transcripts
NM_001171195.1 NM_001171197.1 NM_004432.3 XM_005251393.3 XM_005251394.3 XM_005251395.3 XM_006716734.3 XM_006716735.3 XM_006716736.3 XM_011517774.2 XM_011517776.2 XM_011517777.2 XM_011517778.2 XM_011517779.2 XM_011517780.2 XM_011517783.2 XM_011517784.2 XM_011517785.2 XM_011517786.2 XM_017014408.1 XM_017014409.1 XM_017014410.1 XM_017014411.1 XM_017014412.1 XM_017014413.1 XM_017014414.1 XM_017014415.1 XM_017014416.1 XM_017014417.1 XM_017014418.1 XM_017014419.1 XM_017014420.1 XM_017014421.1 XM_017014422.1 XM_017014423.1 XM_017014424.1 XM_017014425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3906668 c.219C>G p.Y73* Chr.9: 23762100-23762100 on Build GRCh38
4434369 c.149A>C p.N50T Chr.9: 23762170-23762170 on Build GRCh38
1624875 c.54A>G p.P18P Chr.9: 23762181-23762181 on Build GRCh38
3906664 c.232A>G p.M78V Chr.9: 23762087-23762087 on Build GRCh38
3092449 c.60C>T p.T20T Chr.9: 23762175-23762175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754002382 Chr.9: 23762217-23762217 on Build GRCh38
rs745417152 Chr.9: 23762266-23762266 on Build GRCh38
rs577004302 Chr.9: 23762121-23762121 on Build GRCh38
rs750681647 Chr.9: 23762203-23762203 on Build GRCh38
rs73654364 Chr.9: 23762178-23762178 on Build GRCh38
rs973982451 Chr.9: 23762090-23762090 on Build GRCh38
rs760120224 Chr.9: 23762163-23762163 on Build GRCh38
rs766446303 Chr.9: 23762261-23762261 on Build GRCh38
rs76025658 Chr.9: 23762180-23762180 on Build GRCh38
rs756428913 Chr.9: 23762204-23762204 on Build GRCh38
rs762204821 Chr.9: 23762284-23762284 on Build GRCh38
rs758201588 Chr.9: 23762239-23762239 on Build GRCh38
rs775975357 Chr.9: 23762262-23762262 on Build GRCh38
rs1050524983 Chr.9: 23762250-23762250 on Build GRCh38
rs770673494 Chr.9: 23762261-23762261 on Build GRCh38
rs778548697 Chr.9: 23762153-23762153 on Build GRCh38
rs932045794 Chr.9: 23762244-23762244 on Build GRCh38
rs996615141 Chr.9: 23762135-23762135 on Build GRCh38
rs778937550 Chr.9: 23762224-23762224 on Build GRCh38
rs371106420 Chr.9: 23762273-23762273 on Build GRCh38
rs141278970 Chr.9: 23762106-23762106 on Build GRCh38
rs746658089 Chr.9: 23762257-23762257 on Build GRCh38
rs143705199 Chr.9: 23762127-23762127 on Build GRCh38
rs571849626 Chr.9: 23762128-23762128 on Build GRCh38
rs201050464 Chr.9: 23762159-23762159 on Build GRCh38
rs146832864 Chr.9: 23762158-23762158 on Build GRCh38
rs779978734 Chr.9: 23762132-23762132 on Build GRCh38
rs764703332 Chr.9: 23762187-23762187 on Build GRCh38
rs756279278 Chr.9: 23762129-23762129 on Build GRCh38
rs762290569 Chr.9: 23762193-23762193 on Build GRCh38
rs142065322 Chr.9: 23762223-23762223 on Build GRCh38
rs752020952 Chr.9: 23762115-23762115 on Build GRCh38
rs774818542 Chr.9: 23762192-23762192 on Build GRCh38
rs766358082 Chr.9: 23762209-23762209 on Build GRCh38
rs547120448 Chr.9: 23762200-23762200 on Build GRCh38
rs777339981 Chr.9: 23762240-23762240 on Build GRCh38
rs1031950389 Chr.9: 23762259-23762259 on Build GRCh38
rs971054370 Chr.9: 23762164-23762164 on Build GRCh38
rs749180793 Chr.9: 23762134-23762134 on Build GRCh38
rs775012815 Chr.9: 23762281-23762281 on Build GRCh38
rs1008909139 Chr.9: 23762270-23762270 on Build GRCh38
rs770351813 Chr.9: 23762166-23762166 on Build GRCh38
rs747885923 Chr.9: 23762237-23762237 on Build GRCh38
rs935812481 Chr.9: 23762202-23762202 on Build GRCh38
rs768699756 Chr.9: 23762150-23762150 on Build GRCh38

View Full Product Details