Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64340740-64340987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88B
Entrez Gene ID
283234
Gene Name
coiled-coil domain containing 88B
Gene Aliases
BRLZ, CCDC88, HKRP3, gipie
UniGene
Hs.98564
Gene Chromosome Location
Chr.11: 64340203-64357534 on Build GRCh38
Transcripts
NM_032251.5 XM_006718519.3 XM_011544957.2 XM_011544959.2 XM_011544960.2 XM_017017601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5590095 c.275G>A p.W92* Chr.11: 64340975-64340975 on Build GRCh38
6622593 c.214A>G p.S72G Chr.11: 64340914-64340914 on Build GRCh38
5578253 c.218C>T p.S73F Chr.11: 64340918-64340918 on Build GRCh38
4602194 c.286C>T p.H96Y Chr.11: 64340986-64340986 on Build GRCh38
6871246 c.254G>T p.G85V Chr.11: 64340954-64340954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914508831 Chr.11: 64340861-64340861 on Build GRCh38
rs565722976 Chr.11: 64340770-64340770 on Build GRCh38
rs749329571 Chr.11: 64340943-64340943 on Build GRCh38
rs746948276 Chr.11: 64340903-64340903 on Build GRCh38
rs758346668 Chr.11: 64340894-64340894 on Build GRCh38
rs767989966 Chr.11: 64340870-64340870 on Build GRCh38
rs991956743 Chr.11: 64340791-64340791 on Build GRCh38
rs777623406 Chr.11: 64340899-64340899 on Build GRCh38
rs955386975 Chr.11: 64340815-64340815 on Build GRCh38
rs746751891 Chr.11: 64340771-64340771 on Build GRCh38
rs201753153 Chr.11: 64340765-64340765 on Build GRCh38
rs778465444 Chr.11: 64340761-64340761 on Build GRCh38
rs753491110 Chr.11: 64340877-64340877 on Build GRCh38
rs762396613 Chr.11: 64340960-64340960 on Build GRCh38
rs760113917 Chr.11: 64340763-64340764 on Build GRCh38
rs909398066 Chr.11: 64340816-64340816 on Build GRCh38
rs994196884 Chr.11: 64340807-64340807 on Build GRCh38
rs751984259 Chr.11: 64340803-64340803 on Build GRCh38
rs768034960 Chr.11: 64340964-64340964 on Build GRCh38
rs376690430 Chr.11: 64340920-64340920 on Build GRCh38
rs769809463 Chr.11: 64340792-64340792 on Build GRCh38
rs367565462 Chr.11: 64340885-64340885 on Build GRCh38
rs762200392 Chr.11: 64340869-64340869 on Build GRCh38
rs1022274214 Chr.11: 64340921-64340921 on Build GRCh38
rs71456309 Chr.11: 64340790-64340790 on Build GRCh38
rs775452262 Chr.11: 64340923-64340923 on Build GRCh38
rs368470572 Chr.11: 64340775-64340775 on Build GRCh38
rs773587079 Chr.11: 64340968-64340968 on Build GRCh38
rs536590421 Chr.11: 64340786-64340786 on Build GRCh38
rs376856276 Chr.11: 64340852-64340852 on Build GRCh38
rs967671316 Chr.11: 64340879-64340879 on Build GRCh38
rs370077659 Chr.11: 64340952-64340952 on Build GRCh38
rs76402578 Chr.11: 64340802-64340802 on Build GRCh38
rs35027909 Chr.11: 64340897-64340898 on Build GRCh38
rs999162710 Chr.11: 64340944-64340944 on Build GRCh38
rs933494470 Chr.11: 64340854-64340854 on Build GRCh38
rs775604940 Chr.11: 64340797-64340797 on Build GRCh38
rs200730691 Chr.11: 64340826-64340826 on Build GRCh38
rs764685845 Chr.11: 64340891-64340891 on Build GRCh38
rs928314285 Chr.11: 64340959-64340959 on Build GRCh38
rs752203800 Chr.11: 64340893-64340893 on Build GRCh38
rs909238229 Chr.11: 64340827-64340827 on Build GRCh38
rs963855792 Chr.11: 64340850-64340850 on Build GRCh38
rs769289680 Chr.11: 64340947-64340947 on Build GRCh38
rs544170458 Chr.11: 64340800-64340800 on Build GRCh38
rs1050595355 Chr.11: 64340897-64340897 on Build GRCh38
rs780938929 Chr.11: 64340916-64340916 on Build GRCh38
rs772500937 Chr.11: 64340810-64340810 on Build GRCh38
rs1011858732 Chr.11: 64340902-64340902 on Build GRCh38
rs748076825 Chr.11: 64340764-64340764 on Build GRCh38
rs757105963 Chr.11: 64340907-64340907 on Build GRCh38
rs977890339 Chr.11: 64340781-64340781 on Build GRCh38
rs371685182 Chr.11: 64340898-64340898 on Build GRCh38

View Full Product Details