Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30108356-30108611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD3
Entrez Gene ID
79153
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
GDE7
UniGene
Hs.289015
Gene Chromosome Location
Chr.16: 30104810-30113557 on Build GRCh38
Transcripts
NM_024307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3508728 c.762G>A p.S254S Chr.16: 30108378-30108378 on Build GRCh38
3111759 c.761C>T p.S254L Chr.16: 30108379-30108379 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548828892 Chr.16: 30108537-30108537 on Build GRCh38
rs145295505 Chr.16: 30108378-30108378 on Build GRCh38
rs772541406 Chr.16: 30108422-30108422 on Build GRCh38
rs775600369 Chr.16: 30108427-30108427 on Build GRCh38
rs1009998467 Chr.16: 30108483-30108483 on Build GRCh38
rs573744667 Chr.16: 30108509-30108509 on Build GRCh38
rs769478903 Chr.16: 30108467-30108467 on Build GRCh38
rs760700493 Chr.16: 30108447-30108447 on Build GRCh38
rs747808836 Chr.16: 30108463-30108463 on Build GRCh38
rs371158265 Chr.16: 30108429-30108429 on Build GRCh38
rs146701553 Chr.16: 30108383-30108383 on Build GRCh38
rs61764621 Chr.16: 30108503-30108503 on Build GRCh38
rs958893195 Chr.16: 30108528-30108528 on Build GRCh38
rs774161542 Chr.16: 30108400-30108400 on Build GRCh38
rs1035575137 Chr.16: 30108527-30108527 on Build GRCh38
rs746214154 Chr.16: 30108474-30108474 on Build GRCh38
rs764999473 Chr.16: 30108452-30108452 on Build GRCh38
rs754467760 Chr.16: 30108460-30108460 on Build GRCh38
rs577723077 Chr.16: 30108529-30108529 on Build GRCh38
rs982535108 Chr.16: 30108550-30108550 on Build GRCh38
rs751144019 Chr.16: 30108459-30108459 on Build GRCh38
rs61764622 Chr.16: 30108451-30108451 on Build GRCh38
rs780666211 Chr.16: 30108379-30108379 on Build GRCh38
rs746419471 Chr.16: 30108565-30108565 on Build GRCh38
rs780907898 Chr.16: 30108461-30108461 on Build GRCh38
rs760025483 Chr.16: 30108415-30108415 on Build GRCh38
rs774760467 Chr.16: 30108401-30108401 on Build GRCh38
rs374869950 Chr.16: 30108508-30108510 on Build GRCh38
rs374555890 Chr.16: 30108438-30108438 on Build GRCh38
rs982884408 Chr.16: 30108546-30108546 on Build GRCh38
rs375543459 Chr.16: 30108455-30108455 on Build GRCh38
rs534553032 Chr.16: 30108511-30108511 on Build GRCh38
rs893211089 Chr.16: 30108505-30108505 on Build GRCh38
rs780091008 Chr.16: 30108458-30108458 on Build GRCh38
rs779189218 Chr.16: 30108470-30108470 on Build GRCh38
rs1026637525 Chr.16: 30108535-30108535 on Build GRCh38
rs772592660 Chr.16: 30108476-30108476 on Build GRCh38
rs764333654 Chr.16: 30108449-30108449 on Build GRCh38
rs749664859 Chr.16: 30108380-30108380 on Build GRCh38
rs903050101 Chr.16: 30108518-30108518 on Build GRCh38
rs553000395 Chr.16: 30108524-30108524 on Build GRCh38

View Full Product Details