Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137435237-137435498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD8
Entrez Gene ID
377841
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492
UniGene
Hs.512562
Gene Chromosome Location
Chr.9: 137434364-137441710 on Build GRCh38
Transcripts
NM_001033113.1 NM_198585.2 XM_011518665.2 XM_011518666.2 XM_011518668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461408 c.1087C>T p.R363W Chr.9: 137435302-137435302 on Build GRCh38
6654355 c.1083G>A p.W361* Chr.9: 137435306-137435306 on Build GRCh38
1756071 c.1189C>T p.R397C Chr.9: 137435311-137435311 on Build GRCh38
3906065 c.1145T>G p.F382C Chr.9: 137435244-137435244 on Build GRCh38
3906064 c.1256T>G p.F419C Chr.9: 137435244-137435244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202080880 Chr.9: 137435258-137435258 on Build GRCh38
rs140353416 Chr.9: 137435257-137435257 on Build GRCh38
rs530607873 Chr.9: 137435400-137435400 on Build GRCh38
rs1036987754 Chr.9: 137435319-137435319 on Build GRCh38
rs969380977 Chr.9: 137435433-137435435 on Build GRCh38
rs771117470 Chr.9: 137435275-137435275 on Build GRCh38
rs749872587 Chr.9: 137435390-137435390 on Build GRCh38
rs879454321 Chr.9: 137435392-137435392 on Build GRCh38
rs746343980 Chr.9: 137435293-137435293 on Build GRCh38
rs200307848 Chr.9: 137435373-137435373 on Build GRCh38
rs753529973 Chr.9: 137435262-137435262 on Build GRCh38
rs1023338307 Chr.9: 137435310-137435310 on Build GRCh38
rs73565586 Chr.9: 137435440-137435440 on Build GRCh38
rs1012580613 Chr.9: 137435304-137435304 on Build GRCh38
rs769068055 Chr.9: 137435302-137435302 on Build GRCh38
rs781593375 Chr.9: 137435363-137435363 on Build GRCh38
rs574712118 Chr.9: 137435268-137435268 on Build GRCh38
rs563105010 Chr.9: 137435399-137435399 on Build GRCh38
rs535828564 Chr.9: 137435428-137435428 on Build GRCh38
rs78674871 Chr.9: 137435301-137435301 on Build GRCh38
rs567378953 Chr.9: 137435409-137435409 on Build GRCh38
rs943405928 Chr.9: 137435385-137435385 on Build GRCh38
rs550662960 Chr.9: 137435406-137435406 on Build GRCh38
rs933590089 Chr.9: 137435407-137435407 on Build GRCh38
rs546587842 Chr.9: 137435429-137435429 on Build GRCh38
rs770383661 Chr.9: 137435299-137435299 on Build GRCh38
rs760203966 Chr.9: 137435336-137435336 on Build GRCh38
rs753823470 Chr.9: 137435344-137435344 on Build GRCh38
rs751804841 Chr.9: 137435355-137435355 on Build GRCh38
rs1042588470 Chr.9: 137435417-137435417 on Build GRCh38
rs773862600 Chr.9: 137435307-137435307 on Build GRCh38
rs754819736 Chr.9: 137435267-137435267 on Build GRCh38
rs745893259 Chr.9: 137435367-137435367 on Build GRCh38
rs765134519 Chr.9: 137435345-137435345 on Build GRCh38
rs757590818 Chr.9: 137435358-137435358 on Build GRCh38
rs950503997 Chr.9: 137435362-137435362 on Build GRCh38
rs981317208 Chr.9: 137435331-137435331 on Build GRCh38
rs761502158 Chr.9: 137435311-137435311 on Build GRCh38
rs898145311 Chr.9: 137435326-137435326 on Build GRCh38
rs766172009 Chr.9: 137435338-137435338 on Build GRCh38
rs747298693 Chr.9: 137435272-137435272 on Build GRCh38
rs775683857 Chr.9: 137435383-137435383 on Build GRCh38
rs1054651239 Chr.9: 137435451-137435451 on Build GRCh38
rs1029419862 Chr.9: 137435448-137435448 on Build GRCh38
rs572829352 Chr.9: 137435393-137435393 on Build GRCh38
rs560582680 Chr.9: 137435332-137435332 on Build GRCh38
rs1044474353 Chr.9: 137435384-137435385 on Build GRCh38
rs538571885 Chr.9: 137435474-137435474 on Build GRCh38
rs772670458 Chr.9: 137435335-137435335 on Build GRCh38

View Full Product Details