Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 831097-831334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2B
Entrez Gene ID
283229
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
UniGene
Hs.660936
Gene Chromosome Location
Chr.11: 825558-831991 on Build GRCh38
Transcripts
NM_001286606.1 NM_173584.4 XM_011520011.1 XM_011520019.1 XM_017017581.1 XM_017017582.1 XM_017017583.1 XM_017017584.1 XM_017017585.1 XM_017017586.1 XM_017017587.1 XM_017017588.1 XM_017017589.1 XM_017017590.1 XM_017017591.1 XM_017017592.1 XM_017017593.1 XM_017017594.1 XM_017017595.1 XM_017017596.1 XM_017017597.1 XM_017017598.1 XM_017017599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5839709 c.858+1G>A p.? Chr.11: 831296-831296 on Build GRCh38
4146381 c.798C>A p.R266R Chr.11: 831235-831235 on Build GRCh38
6895135 c.829C>T p.Q277* Chr.11: 831266-831266 on Build GRCh38
4680290 c.838A>G p.T280A Chr.11: 831275-831275 on Build GRCh38
2220743 c.831G>C p.Q277H Chr.11: 831268-831268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778313411 Chr.11: 831299-831299 on Build GRCh38
rs777799390 Chr.11: 831233-831233 on Build GRCh38
rs758880815 Chr.11: 831228-831228 on Build GRCh38
rs753043393 Chr.11: 831291-831291 on Build GRCh38
rs760422964 Chr.11: 831116-831116 on Build GRCh38
rs752139841 Chr.11: 831135-831135 on Build GRCh38
rs769664886 Chr.11: 831251-831251 on Build GRCh38
rs1003672704 Chr.11: 831128-831128 on Build GRCh38
rs749706588 Chr.11: 831152-831152 on Build GRCh38
rs765289896 Chr.11: 831131-831131 on Build GRCh38
rs756495724 Chr.11: 831144-831144 on Build GRCh38
rs776459130 Chr.11: 831120-831120 on Build GRCh38
rs779532396 Chr.11: 831157-831157 on Build GRCh38
rs781355491 Chr.11: 831306-831306 on Build GRCh38
rs779219869 Chr.11: 831219-831219 on Build GRCh38
rs373031263 Chr.11: 831276-831276 on Build GRCh38
rs747160077 Chr.11: 831237-831237 on Build GRCh38
rs768514186 Chr.11: 831258-831258 on Build GRCh38
rs199501864 Chr.11: 831250-831250 on Build GRCh38
rs755534481 Chr.11: 831236-831236 on Build GRCh38
rs550772293 Chr.11: 831143-831143 on Build GRCh38
rs759777966 Chr.11: 831277-831277 on Build GRCh38
rs376369663 Chr.11: 831191-831191 on Build GRCh38
rs7114218 Chr.11: 831122-831122 on Build GRCh38
rs771183379 Chr.11: 831173-831173 on Build GRCh38
rs757941557 Chr.11: 831138-831138 on Build GRCh38
rs746121627 Chr.11: 831315-831315 on Build GRCh38
rs191318331 Chr.11: 831205-831205 on Build GRCh38
rs761465228 Chr.11: 831269-831269 on Build GRCh38
rs753270877 Chr.11: 831227-831227 on Build GRCh38
rs772196202 Chr.11: 831163-831163 on Build GRCh38
rs775238953 Chr.11: 831253-831253 on Build GRCh38
rs12222600 Chr.11: 831311-831311 on Build GRCh38
rs368929641 Chr.11: 831270-831270 on Build GRCh38
rs758941366 Chr.11: 831298-831298 on Build GRCh38
rs763224147 Chr.11: 831257-831257 on Build GRCh38
rs536436212 Chr.11: 831268-831268 on Build GRCh38
rs754240879 Chr.11: 831211-831211 on Build GRCh38
rs34889510 Chr.11: 831316-831316 on Build GRCh38
rs751160179 Chr.11: 831196-831196 on Build GRCh38
rs868797816 Chr.11: 831139-831139 on Build GRCh38
rs373514484 Chr.11: 831245-831245 on Build GRCh38
rs765029739 Chr.11: 831184-831184 on Build GRCh38
rs762921111 Chr.11: 831254-831254 on Build GRCh38
rs369206850 Chr.11: 831199-831199 on Build GRCh38
rs755434931 Chr.11: 831154-831154 on Build GRCh38
rs1034782125 Chr.11: 831142-831142 on Build GRCh38
rs965779966 Chr.11: 831260-831260 on Build GRCh38
rs747211542 Chr.11: 831166-831166 on Build GRCh38
rs748117705 Chr.11: 831159-831159 on Build GRCh38
rs781410525 Chr.11: 831249-831249 on Build GRCh38
rs780540563 Chr.11: 831150-831150 on Build GRCh38
rs763680224 Chr.11: 831193-831193 on Build GRCh38
rs749815557 Chr.11: 831179-831179 on Build GRCh38
rs34978511 Chr.11: 831187-831188 on Build GRCh38
rs201736358 Chr.11: 831183-831183 on Build GRCh38
rs751584263 Chr.11: 831300-831300 on Build GRCh38
rs766672004 Chr.11: 831287-831287 on Build GRCh38
rs765613477 Chr.11: 831281-831281 on Build GRCh38
rs776409856 Chr.11: 831178-831178 on Build GRCh38
rs762326715 Chr.11: 831192-831192 on Build GRCh38

View Full Product Details