Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 190225909-190226170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP3
Entrez Gene ID
339479
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
DBCCR1L, DBCCR1L1, FAM5C
UniGene
Hs.65765
Gene Chromosome Location
Chr.1: 190097662-190477997 on Build GRCh38
Transcripts
NM_001317188.1 NM_199051.2 XM_011509472.2 XM_011509475.2 XM_011509476.2 XM_017001125.1 XM_017001126.1 XM_017001127.1 XM_017001128.1 XM_017001129.1 XM_017001130.1 XM_017001131.1 XM_017001132.1 XM_017001133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700559 c.961+2T>C p.? Chr.1: 190226080-190226080 on Build GRCh38
1206401 c.893C>T p.A298V Chr.1: 190226150-190226150 on Build GRCh38
6307248 c.899A>T p.E300V Chr.1: 190226144-190226144 on Build GRCh38
1172080 c.908T>G p.L303R Chr.1: 190226135-190226135 on Build GRCh38
677453 c.928T>A p.W310R Chr.1: 190226115-190226115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147573045 Chr.1: 190226148-190226148 on Build GRCh38
rs769015923 Chr.1: 190226079-190226079 on Build GRCh38
rs780316120 Chr.1: 190226074-190226074 on Build GRCh38
rs372241341 Chr.1: 190226064-190226064 on Build GRCh38
rs997044320 Chr.1: 190225967-190225967 on Build GRCh38
rs199613980 Chr.1: 190226109-190226109 on Build GRCh38
rs891280238 Chr.1: 190226012-190226012 on Build GRCh38
rs770672623 Chr.1: 190226098-190226098 on Build GRCh38
rs757626264 Chr.1: 190226059-190226059 on Build GRCh38
rs990520433 Chr.1: 190226034-190226034 on Build GRCh38
rs748399068 Chr.1: 190226094-190226094 on Build GRCh38
rs763653559 Chr.1: 190226135-190226135 on Build GRCh38
rs774992892 Chr.1: 190226129-190226129 on Build GRCh38
rs767792284 Chr.1: 190226150-190226150 on Build GRCh38
rs374344704 Chr.1: 190226075-190226075 on Build GRCh38
rs200983517 Chr.1: 190226123-190226123 on Build GRCh38
rs957424967 Chr.1: 190226024-190226024 on Build GRCh38
rs769652831 Chr.1: 190226113-190226113 on Build GRCh38
rs113747384 Chr.1: 190225952-190225952 on Build GRCh38
rs536217568 Chr.1: 190225956-190225956 on Build GRCh38
rs998918760 Chr.1: 190226020-190226020 on Build GRCh38
rs779991607 Chr.1: 190226056-190226058 on Build GRCh38
rs1033065182 Chr.1: 190226035-190226036 on Build GRCh38
rs752116455 Chr.1: 190226061-190226068 on Build GRCh38
rs1029755657 Chr.1: 190225979-190225979 on Build GRCh38
rs200969183 Chr.1: 190226062-190226062 on Build GRCh38
rs751208820 Chr.1: 190226139-190226139 on Build GRCh38
rs762506239 Chr.1: 190226132-190226132 on Build GRCh38
rs367704040 Chr.1: 190226106-190226106 on Build GRCh38
rs756644682 Chr.1: 190226072-190226072 on Build GRCh38
rs779063923 Chr.1: 190226089-190226089 on Build GRCh38
rs865794022 Chr.1: 190226142-190226142 on Build GRCh38
rs140670727 Chr.1: 190226035-190226035 on Build GRCh38
rs750588100 Chr.1: 190226036-190226036 on Build GRCh38
rs1031714359 Chr.1: 190226021-190226021 on Build GRCh38
rs553396217 Chr.1: 190225992-190225992 on Build GRCh38
rs900045294 Chr.1: 190225966-190225966 on Build GRCh38
rs758544826 Chr.1: 190226054-190226055 on Build GRCh38

View Full Product Details