Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155026351-155026602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
UniGene
Hs.591491
Gene Chromosome Location
Chr.1: 155018513-155033781 on Build GRCh38
Transcripts
NM_144622.2 XM_011509188.2 XM_011509189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574408 c.1558G>C p.V520L Chr.1: 155026355-155026355 on Build GRCh38
3930417 c.1459C>A p.R487S Chr.1: 155026599-155026599 on Build GRCh38
6641052 c.1554C>A p.S518R Chr.1: 155026359-155026359 on Build GRCh38
280322 c.1523G>A p.G508D Chr.1: 155026390-155026390 on Build GRCh38
1334521 c.1472G>A p.R491H Chr.1: 155026586-155026586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765092835 Chr.1: 155026550-155026550 on Build GRCh38
rs767432489 Chr.1: 155026529-155026529 on Build GRCh38
rs924679307 Chr.1: 155026574-155026574 on Build GRCh38
rs749108165 Chr.1: 155026400-155026400 on Build GRCh38
rs756052451 Chr.1: 155026396-155026396 on Build GRCh38
rs372734773 Chr.1: 155026511-155026511 on Build GRCh38
rs762349954 Chr.1: 155026516-155026516 on Build GRCh38
rs774903744 Chr.1: 155026429-155026429 on Build GRCh38
rs557270335 Chr.1: 155026517-155026517 on Build GRCh38
rs998080784 Chr.1: 155026496-155026496 on Build GRCh38
rs952544308 Chr.1: 155026483-155026483 on Build GRCh38
rs897608921 Chr.1: 155026486-155026486 on Build GRCh38
rs758642669 Chr.1: 155026374-155026374 on Build GRCh38
rs972358701 Chr.1: 155026466-155026466 on Build GRCh38
rs977458312 Chr.1: 155026551-155026551 on Build GRCh38
rs201166423 Chr.1: 155026535-155026535 on Build GRCh38
rs773668079 Chr.1: 155026518-155026518 on Build GRCh38
rs138732547 Chr.1: 155026559-155026559 on Build GRCh38
rs751644853 Chr.1: 155026523-155026523 on Build GRCh38
rs745542659 Chr.1: 155026421-155026421 on Build GRCh38
rs1005888901 Chr.1: 155026467-155026467 on Build GRCh38
rs777757292 Chr.1: 155026397-155026397 on Build GRCh38
rs746506368 Chr.1: 155026431-155026431 on Build GRCh38
rs781624038 Chr.1: 155026438-155026439 on Build GRCh38
rs887454027 Chr.1: 155026464-155026464 on Build GRCh38
rs748236964 Chr.1: 155026442-155026465 on Build GRCh38
rs1044684999 Chr.1: 155026457-155026457 on Build GRCh38
rs985227170 Chr.1: 155026493-155026493 on Build GRCh38
rs369622293 Chr.1: 155026465-155026465 on Build GRCh38
rs896377649 Chr.1: 155026401-155026401 on Build GRCh38
rs750188363 Chr.1: 155026569-155026569 on Build GRCh38
rs755233012 Chr.1: 155026433-155026433 on Build GRCh38
rs759661176 Chr.1: 155026525-155026525 on Build GRCh38
rs544633457 Chr.1: 155026476-155026476 on Build GRCh38
rs935265817 Chr.1: 155026571-155026571 on Build GRCh38
rs199571532 Chr.1: 155026580-155026580 on Build GRCh38
rs751339482 Chr.1: 155026402-155026402 on Build GRCh38
rs757138608 Chr.1: 155026555-155026555 on Build GRCh38
rs766398746 Chr.1: 155026375-155026375 on Build GRCh38
rs769109264 Chr.1: 155026437-155026437 on Build GRCh38
rs199908001 Chr.1: 155026557-155026557 on Build GRCh38
rs1035901573 Chr.1: 155026454-155026454 on Build GRCh38
rs751623759 Chr.1: 155026393-155026393 on Build GRCh38
rs771674076 Chr.1: 155026422-155026422 on Build GRCh38
rs369182906 Chr.1: 155026501-155026501 on Build GRCh38
rs758036788 Chr.1: 155026579-155026579 on Build GRCh38
rs201207318 Chr.1: 155026409-155026409 on Build GRCh38
rs878964608 Chr.1: 155026461-155026461 on Build GRCh38
rs778624170 Chr.1: 155026414-155026414 on Build GRCh38
rs763305094 Chr.1: 155026520-155026520 on Build GRCh38

View Full Product Details