Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38293164-38293405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1E
Entrez Gene ID
1454
Gene Name
casein kinase 1 epsilon
Gene Aliases
CKIepsilon, HCKIE
UniGene
Hs.474833
Gene Chromosome Location
Chr.22: 38290691-38318084 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Gene Symbol
LOC400927-CSNK1E
Entrez Gene ID
102800317
Gene Name
LOC400927-CSNK1E readthrough
Gene Aliases
-
UniGene
Hs.720223
Gene Chromosome Location
Chr.22: 38290691-38398522 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4104073 c.1229C>T p.P410L Chr.22: 38293309-38293309 on Build GRCh38
1559606 c.1243G>A p.G415R Chr.22: 38293295-38293295 on Build GRCh38
1559605 c.1243G>A p.G415R Chr.22: 38293295-38293295 on Build GRCh38
4104074 c.1229C>T p.P410L Chr.22: 38293309-38293309 on Build GRCh38
4104072 c.1229C>T p.P410L Chr.22: 38293309-38293309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35665927 Chr.22: 38293300-38293300 on Build GRCh38
rs760485182 Chr.22: 38293368-38293368 on Build GRCh38
rs536877806 Chr.22: 38293350-38293350 on Build GRCh38
rs893439075 Chr.22: 38293244-38293244 on Build GRCh38
rs756000157 Chr.22: 38293295-38293295 on Build GRCh38
rs532233715 Chr.22: 38293296-38293296 on Build GRCh38
rs755994792 Chr.22: 38293331-38293344 on Build GRCh38
rs769102255 Chr.22: 38293247-38293247 on Build GRCh38
rs762304380 Chr.22: 38293265-38293265 on Build GRCh38
rs775282864 Chr.22: 38293362-38293362 on Build GRCh38
rs1015504687 Chr.22: 38293384-38293384 on Build GRCh38
rs569420536 Chr.22: 38293334-38293334 on Build GRCh38
rs142283293 Chr.22: 38293275-38293275 on Build GRCh38
rs910487898 Chr.22: 38293304-38293304 on Build GRCh38
rs374879706 Chr.22: 38293356-38293356 on Build GRCh38
rs767327253 Chr.22: 38293276-38293276 on Build GRCh38
rs371648738 Chr.22: 38293336-38293336 on Build GRCh38
rs1049407389 Chr.22: 38293232-38293232 on Build GRCh38
rs904717263 Chr.22: 38293386-38293386 on Build GRCh38
rs377722777 Chr.22: 38293337-38293337 on Build GRCh38
rs775804860 Chr.22: 38293238-38293238 on Build GRCh38
rs775203471 Chr.22: 38293302-38293302 on Build GRCh38
rs368611889 Chr.22: 38293348-38293348 on Build GRCh38
rs758990683 Chr.22: 38293357-38293357 on Build GRCh38
rs747423925 Chr.22: 38293246-38293246 on Build GRCh38
rs769014406 Chr.22: 38293345-38293345 on Build GRCh38
rs77945315 Chr.22: 38293315-38293315 on Build GRCh38
rs757203185 Chr.22: 38293309-38293309 on Build GRCh38
rs185497677 Chr.22: 38293216-38293216 on Build GRCh38
rs1027941531 Chr.22: 38293283-38293283 on Build GRCh38
rs952004020 Chr.22: 38293347-38293347 on Build GRCh38
rs1013896567 Chr.22: 38293268-38293268 on Build GRCh38
rs201579666 Chr.22: 38293278-38293278 on Build GRCh38
rs745675429 Chr.22: 38293330-38293343 on Build GRCh38
rs779048216 Chr.22: 38293311-38293311 on Build GRCh38
rs371135313 Chr.22: 38293274-38293274 on Build GRCh38
rs55755354 Chr.22: 38293273-38293273 on Build GRCh38
rs779707543 Chr.22: 38293332-38293335 on Build GRCh38
rs370427643 Chr.22: 38293354-38293354 on Build GRCh38
rs367551605 Chr.22: 38293358-38293358 on Build GRCh38
rs777155984 Chr.22: 38293252-38293252 on Build GRCh38
rs747334057 Chr.22: 38293338-38293338 on Build GRCh38
rs760447047 Chr.22: 38293373-38293373 on Build GRCh38
rs894106618 Chr.22: 38293217-38293217 on Build GRCh38
rs1012714708 Chr.22: 38293313-38293313 on Build GRCh38

View Full Product Details