Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151955507-151955752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRE
Entrez Gene ID
2564
Gene Name
gamma-aminobutyric acid type A receptor epsilon subunit
Gene Aliases
-
UniGene
Hs.22785
Gene Chromosome Location
Chr.X: 151953124-151974679 on Build GRCh38
Transcripts
NM_004961.3 XM_011531140.2 XM_017029386.1 XM_017029387.1 XM_017029388.1 XM_017029389.1

SNP Detail

SNP ID Chromosome Location
rs773206156 Chr.X: 151955596-151955604 on Build GRCh38
rs374571425 Chr.X: 151955597-151955597 on Build GRCh38
rs779576822 Chr.X: 151955563-151955563 on Build GRCh38
rs77007790 Chr.X: 151955549-151955549 on Build GRCh38
rs936314042 Chr.X: 151955551-151955551 on Build GRCh38
rs147982495 Chr.X: 151955672-151955672 on Build GRCh38
rs78212023 Chr.X: 151955566-151955566 on Build GRCh38
rs367673721 Chr.X: 151955685-151955685 on Build GRCh38
rs776156895 Chr.X: 151955660-151955660 on Build GRCh38
rs749496766 Chr.X: 151955570-151955570 on Build GRCh38
rs778197680 Chr.X: 151955616-151955616 on Build GRCh38
rs759119898 Chr.X: 151955662-151955662 on Build GRCh38
rs924907083 Chr.X: 151955539-151955539 on Build GRCh38
rs951985435 Chr.X: 151955679-151955679 on Build GRCh38
rs483352728 Chr.X: 151955530-151955530 on Build GRCh38
rs768865712 Chr.X: 151955579-151955579 on Build GRCh38
rs187011945 Chr.X: 151955686-151955686 on Build GRCh38
rs112646382 Chr.X: 151955538-151955538 on Build GRCh38
rs751530972 Chr.X: 151955689-151955689 on Build GRCh38
rs753889158 Chr.X: 151955726-151955726 on Build GRCh38
rs371442646 Chr.X: 151955619-151955619 on Build GRCh38
rs766386512 Chr.X: 151955719-151955719 on Build GRCh38
rs745335766 Chr.X: 151955702-151955702 on Build GRCh38
rs753074937 Chr.X: 151955558-151955558 on Build GRCh38
rs756914368 Chr.X: 151955675-151955675 on Build GRCh38
rs181697394 Chr.X: 151955612-151955612 on Build GRCh38
rs775117386 Chr.X: 151955674-151955674 on Build GRCh38
rs896303707 Chr.X: 151955661-151955661 on Build GRCh38
rs748590514 Chr.X: 151955602-151955602 on Build GRCh38
rs201300022 Chr.X: 151955586-151955586 on Build GRCh38
rs772552823 Chr.X: 151955609-151955609 on Build GRCh38

View Full Product Details