Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3643662-3643935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC11
Entrez Gene ID
78989
Gene Name
collectin subfamily member 11
Gene Aliases
3MC2, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, CLK1
UniGene
Hs.32603
Gene Chromosome Location
Chr.2: 3594832-3644644 on Build GRCh38
Transcripts
NM_001255982.1 NM_001255983.1 NM_001255984.1 NM_001255985.1 NM_001255986.1 NM_001255987.1 NM_001255988.1 NM_001255989.1 NM_024027.4 NM_199235.2 NR_045659.1 XM_005263853.4 XM_006711897.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5564268 c.430C>T p.R144C Chr.2: 3643741-3643741 on Build GRCh38
1668685 c.641G>A p.R214H Chr.2: 3643901-3643901 on Build GRCh38
4094029 c.482C>T p.A161V Chr.2: 3643793-3643793 on Build GRCh38
5603268 c.606C>T p.I202I Chr.2: 3643917-3643917 on Build GRCh38
4094030 c.533C>T p.A178V Chr.2: 3643793-3643793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759573402 Chr.2: 3643780-3643780 on Build GRCh38
rs1047598580 Chr.2: 3643812-3643812 on Build GRCh38
rs763926251 Chr.2: 3643875-3643875 on Build GRCh38
rs777019867 Chr.2: 3643759-3643759 on Build GRCh38
rs761940723 Chr.2: 3643764-3643764 on Build GRCh38
rs374563577 Chr.2: 3643724-3643724 on Build GRCh38
rs772874935 Chr.2: 3643776-3643776 on Build GRCh38
rs746438566 Chr.2: 3643742-3643742 on Build GRCh38
rs964446994 Chr.2: 3643713-3643713 on Build GRCh38
rs760432690 Chr.2: 3643866-3643866 on Build GRCh38
rs199526207 Chr.2: 3643767-3643767 on Build GRCh38
rs387907075 Chr.2: 3643807-3643807 on Build GRCh38
rs113757131 Chr.2: 3643858-3643858 on Build GRCh38
rs896580554 Chr.2: 3643793-3643793 on Build GRCh38
rs768946186 Chr.2: 3643753-3643753 on Build GRCh38
rs773407043 Chr.2: 3643830-3643830 on Build GRCh38
rs267599363 Chr.2: 3643809-3643809 on Build GRCh38
rs545129835 Chr.2: 3643797-3643797 on Build GRCh38
rs762172005 Chr.2: 3643826-3643826 on Build GRCh38
rs769170798 Chr.2: 3643821-3643821 on Build GRCh38
rs777356067 Chr.2: 3643792-3643792 on Build GRCh38
rs138441935 Chr.2: 3643794-3643794 on Build GRCh38
rs753906527 Chr.2: 3643688-3643688 on Build GRCh38
rs781087377 Chr.2: 3643726-3643726 on Build GRCh38
rs747333394 Chr.2: 3643820-3643820 on Build GRCh38
rs757871477 Chr.2: 3643885-3643885 on Build GRCh38
rs868514360 Chr.2: 3643913-3643913 on Build GRCh38
rs767532219 Chr.2: 3643717-3643718 on Build GRCh38
rs150837001 Chr.2: 3643848-3643848 on Build GRCh38
rs368720161 Chr.2: 3643787-3643787 on Build GRCh38
rs764804437 Chr.2: 3643882-3643882 on Build GRCh38
rs751543343 Chr.2: 3643719-3643719 on Build GRCh38
rs387907076 Chr.2: 3643912-3643912 on Build GRCh38
rs988678931 Chr.2: 3643909-3643909 on Build GRCh38
rs1056975548 Chr.2: 3643791-3643791 on Build GRCh38
rs150077720 Chr.2: 3643727-3643727 on Build GRCh38
rs757428964 Chr.2: 3643711-3643711 on Build GRCh38
rs553743194 Chr.2: 3643878-3643878 on Build GRCh38
rs769978108 Chr.2: 3643829-3643829 on Build GRCh38
rs747554313 Chr.2: 3643891-3643891 on Build GRCh38
rs1017265813 Chr.2: 3643872-3643872 on Build GRCh38
rs760954666 Chr.2: 3643749-3643749 on Build GRCh38
rs141691492 Chr.2: 3643734-3643734 on Build GRCh38
rs572396920 Chr.2: 3643911-3643911 on Build GRCh38
rs374212450 Chr.2: 3643884-3643884 on Build GRCh38
rs577671414 Chr.2: 3643844-3643844 on Build GRCh38
rs767118530 Chr.2: 3643736-3643736 on Build GRCh38
rs367740433 Chr.2: 3643890-3643890 on Build GRCh38
rs139848694 Chr.2: 3643883-3643883 on Build GRCh38
rs767209845 Chr.2: 3643786-3643786 on Build GRCh38
rs748736842 Chr.2: 3643735-3643735 on Build GRCh38
rs866955039 Chr.2: 3643823-3643823 on Build GRCh38
rs755830142 Chr.2: 3643731-3643731 on Build GRCh38
rs772312145 Chr.2: 3643743-3643743 on Build GRCh38
rs775993317 Chr.2: 3643748-3643748 on Build GRCh38
rs779362328 Chr.2: 3643737-3643737 on Build GRCh38
rs866636297 Chr.2: 3643873-3643873 on Build GRCh38
rs988342294 Chr.2: 3643740-3643740 on Build GRCh38
rs148786016 Chr.2: 3643816-3643816 on Build GRCh38
rs776958693 Chr.2: 3643822-3643822 on Build GRCh38
rs766366668 Chr.2: 3643718-3643718 on Build GRCh38
rs751427364 Chr.2: 3643774-3643774 on Build GRCh38
rs780443106 Chr.2: 3643819-3643819 on Build GRCh38
rs765878784 Chr.2: 3643879-3643879 on Build GRCh38

View Full Product Details