Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34852172-34852409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
UniGene
Hs.433738
Gene Chromosome Location
Chr.20: 34844720-34872858 on Build GRCh38
Transcripts
NM_178026.2 XM_011528782.2 XM_011528783.2 XM_017027809.1 XM_017027810.1 XM_017027811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5914293 c.1547C>T p.S516F Chr.20: 34852195-34852195 on Build GRCh38
3545576 c.1474C>T p.L492L Chr.20: 34852268-34852268 on Build GRCh38
6588852 c.1473C>T p.S491S Chr.20: 34852269-34852269 on Build GRCh38
6093326 c.1511C>T p.S504L Chr.20: 34852231-34852231 on Build GRCh38
4686888 c.1471T>C p.S491P Chr.20: 34852271-34852271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537150694 Chr.20: 34852275-34852275 on Build GRCh38
rs1030634700 Chr.20: 34852326-34852326 on Build GRCh38
rs558657735 Chr.20: 34852320-34852320 on Build GRCh38
rs866176443 Chr.20: 34852236-34852236 on Build GRCh38
rs778644849 Chr.20: 34852234-34852234 on Build GRCh38
rs878895916 Chr.20: 34852283-34852283 on Build GRCh38
rs975781895 Chr.20: 34852343-34852343 on Build GRCh38
rs762172271 Chr.20: 34852282-34852282 on Build GRCh38
rs764414645 Chr.20: 34852202-34852202 on Build GRCh38
rs766488910 Chr.20: 34852313-34852313 on Build GRCh38
rs746544534 Chr.20: 34852248-34852248 on Build GRCh38
rs774636144 Chr.20: 34852279-34852279 on Build GRCh38
rs946760966 Chr.20: 34852359-34852359 on Build GRCh38
rs991009167 Chr.20: 34852331-34852331 on Build GRCh38
rs138441861 Chr.20: 34852204-34852204 on Build GRCh38
rs201339481 Chr.20: 34852191-34852191 on Build GRCh38
rs201177862 Chr.20: 34852336-34852336 on Build GRCh38
rs776256348 Chr.20: 34852261-34852261 on Build GRCh38
rs747530699 Chr.20: 34852237-34852237 on Build GRCh38
rs758829060 Chr.20: 34852215-34852215 on Build GRCh38
rs377137760 Chr.20: 34852365-34852365 on Build GRCh38
rs773376870 Chr.20: 34852304-34852304 on Build GRCh38
rs767765280 Chr.20: 34852214-34852214 on Build GRCh38
rs370540483 Chr.20: 34852375-34852375 on Build GRCh38
rs770514344 Chr.20: 34852257-34852257 on Build GRCh38
rs534641558 Chr.20: 34852364-34852364 on Build GRCh38
rs768780046 Chr.20: 34852298-34852299 on Build GRCh38
rs778406384 Chr.20: 34852233-34852233 on Build GRCh38
rs569819094 Chr.20: 34852235-34852235 on Build GRCh38
rs142938362 Chr.20: 34852231-34852231 on Build GRCh38
rs750508603 Chr.20: 34852218-34852218 on Build GRCh38
rs771222124 Chr.20: 34852312-34852312 on Build GRCh38
rs756149488 Chr.20: 34852222-34852222 on Build GRCh38
rs777413502 Chr.20: 34852242-34852242 on Build GRCh38
rs758168191 Chr.20: 34852379-34852379 on Build GRCh38
rs772215890 Chr.20: 34852292-34852292 on Build GRCh38
rs760866289 Chr.20: 34852307-34852307 on Build GRCh38
rs766326804 Chr.20: 34852227-34852227 on Build GRCh38
rs762005863 Chr.20: 34852209-34852209 on Build GRCh38
rs769202495 Chr.20: 34852278-34852278 on Build GRCh38
rs984073016 Chr.20: 34852303-34852303 on Build GRCh38
rs753913102 Chr.20: 34852318-34852318 on Build GRCh38
rs774746188 Chr.20: 34852205-34852205 on Build GRCh38

View Full Product Details