Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43873771-43874044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD5
Entrez Gene ID
84978
Gene Name
FERM domain containing 5
Gene Aliases
-
UniGene
Hs.578544
Gene Chromosome Location
Chr.15: 43870761-44195294 on Build GRCh38
Transcripts
NM_001286490.1 NM_001286491.1 NM_001322949.1 NM_001322950.1 NM_001322951.1 NM_032892.4 NR_104455.1 XM_005254730.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2188865 c.1667T>G p.F556C Chr.15: 43873931-43873931 on Build GRCh38
5730435 c.1632C>A p.F544L Chr.15: 43873966-43873966 on Build GRCh38
2188868 c.1585A>G p.I529V Chr.15: 43874013-43874013 on Build GRCh38
6303067 c.1660C>A p.R554R Chr.15: 43873938-43873938 on Build GRCh38
7316378 c.1596C>T p.F532F Chr.15: 43874002-43874002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373069405 Chr.15: 43873895-43873895 on Build GRCh38
rs760564859 Chr.15: 43873981-43873981 on Build GRCh38
rs751456138 Chr.15: 43873869-43873869 on Build GRCh38
rs889157780 Chr.15: 43873819-43873819 on Build GRCh38
rs372074731 Chr.15: 43873838-43873838 on Build GRCh38
rs915808887 Chr.15: 43874013-43874013 on Build GRCh38
rs753549302 Chr.15: 43873991-43873991 on Build GRCh38
rs752602387 Chr.15: 43873889-43873889 on Build GRCh38
rs746378483 Chr.15: 43873916-43873916 on Build GRCh38
rs369799326 Chr.15: 43873918-43873918 on Build GRCh38
rs752551012 Chr.15: 43873980-43873980 on Build GRCh38
rs936947960 Chr.15: 43873808-43873808 on Build GRCh38
rs768987510 Chr.15: 43873923-43873923 on Build GRCh38
rs541456165 Chr.15: 43873796-43873796 on Build GRCh38
rs780212034 Chr.15: 43874004-43874004 on Build GRCh38
rs369114095 Chr.15: 43873877-43873877 on Build GRCh38
rs376004230 Chr.15: 43873867-43873867 on Build GRCh38
rs747415871 Chr.15: 43873922-43873922 on Build GRCh38
rs756855308 Chr.15: 43873902-43873902 on Build GRCh38
rs376269021 Chr.15: 43873978-43873978 on Build GRCh38
rs768936248 Chr.15: 43873845-43873845 on Build GRCh38
rs140698102 Chr.15: 43873807-43873807 on Build GRCh38
rs773397243 Chr.15: 43873963-43873963 on Build GRCh38
rs769875158 Chr.15: 43873941-43873941 on Build GRCh38
rs763937333 Chr.15: 43873985-43873985 on Build GRCh38
rs755940494 Chr.15: 43873891-43873891 on Build GRCh38
rs564140199 Chr.15: 43873945-43873945 on Build GRCh38
rs111817812 Chr.15: 43874002-43874002 on Build GRCh38
rs761931035 Chr.15: 43873865-43873865 on Build GRCh38
rs778424625 Chr.15: 43873906-43873906 on Build GRCh38
rs548650879 Chr.15: 43874000-43874000 on Build GRCh38
rs762935838 Chr.15: 43873976-43873976 on Build GRCh38
rs761955840 Chr.15: 43873940-43873940 on Build GRCh38
rs867923161 Chr.15: 43873853-43873853 on Build GRCh38
rs757817954 Chr.15: 43874012-43874012 on Build GRCh38
rs150478495 Chr.15: 43873837-43873837 on Build GRCh38
rs776803947 Chr.15: 43873859-43873859 on Build GRCh38
rs1006096376 Chr.15: 43873961-43873961 on Build GRCh38
rs760936531 Chr.15: 43873843-43873843 on Build GRCh38
rs375941965 Chr.15: 43873896-43873896 on Build GRCh38
rs199998801 Chr.15: 43873879-43873879 on Build GRCh38
rs201700276 Chr.15: 43873937-43873937 on Build GRCh38
rs563506964 Chr.15: 43873917-43873917 on Build GRCh38
rs764786323 Chr.15: 43874001-43874001 on Build GRCh38
rs753488164 Chr.15: 43873900-43873900 on Build GRCh38
rs750019572 Chr.15: 43874007-43874007 on Build GRCh38
rs551899362 Chr.15: 43873818-43873818 on Build GRCh38

View Full Product Details