Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 26434194-26434431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPHD2
Entrez Gene ID
57168
Gene Name
aspartate beta-hydroxylase domain containing 2
Gene Aliases
-
UniGene
Hs.567547
Gene Chromosome Location
Chr.22: 26429314-26445012 on Build GRCh38
Transcripts
NM_020437.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4793984 c.560G>A p.S187N Chr.22: 26434253-26434253 on Build GRCh38
2935494 c.755G>A p.R252H Chr.22: 26434370-26434370 on Build GRCh38
7121931 c.615C>T p.L205L Chr.22: 26434230-26434230 on Build GRCh38
1130470 c.727G>A p.A243T Chr.22: 26434420-26434420 on Build GRCh38
2935484 c.587A>G p.Q196R Chr.22: 26434202-26434202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148508323 Chr.22: 26434300-26434300 on Build GRCh38
rs146331448 Chr.22: 26434289-26434289 on Build GRCh38
rs779187230 Chr.22: 26434284-26434284 on Build GRCh38
rs776616723 Chr.22: 26434312-26434312 on Build GRCh38
rs775196043 Chr.22: 26434325-26434325 on Build GRCh38
rs753021769 Chr.22: 26434365-26434365 on Build GRCh38
rs756785653 Chr.22: 26434394-26434394 on Build GRCh38
rs773584115 Chr.22: 26434341-26434341 on Build GRCh38
rs754339838 Chr.22: 26434360-26434360 on Build GRCh38
rs750493648 Chr.22: 26434248-26434248 on Build GRCh38
rs780554272 Chr.22: 26434259-26434259 on Build GRCh38
rs554601838 Chr.22: 26434293-26434293 on Build GRCh38
rs754364586 Chr.22: 26434260-26434260 on Build GRCh38
rs371659241 Chr.22: 26434295-26434295 on Build GRCh38
rs144492202 Chr.22: 26434362-26434362 on Build GRCh38
rs770550097 Chr.22: 26434332-26434332 on Build GRCh38
rs979401849 Chr.22: 26434345-26434345 on Build GRCh38
rs746721354 Chr.22: 26434377-26434377 on Build GRCh38
rs182629777 Chr.22: 26434290-26434290 on Build GRCh38
rs374955523 Chr.22: 26434311-26434311 on Build GRCh38
rs767679005 Chr.22: 26434220-26434220 on Build GRCh38
rs144268277 Chr.22: 26434256-26434256 on Build GRCh38
rs150592332 Chr.22: 26434331-26434331 on Build GRCh38
rs896328520 Chr.22: 26434405-26434405 on Build GRCh38
rs147832707 Chr.22: 26434364-26434364 on Build GRCh38
rs11705277 Chr.22: 26434350-26434350 on Build GRCh38
rs760828431 Chr.22: 26434349-26434349 on Build GRCh38
rs769375246 Chr.22: 26434371-26434371 on Build GRCh38
rs777347826 Chr.22: 26434375-26434375 on Build GRCh38
rs34902186 Chr.22: 26434319-26434319 on Build GRCh38
rs769596988 Chr.22: 26434322-26434322 on Build GRCh38
rs755366536 Chr.22: 26434279-26434279 on Build GRCh38

View Full Product Details