Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200817849-200818122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BZW1
Entrez Gene ID
9689
Gene Name
basic leucine zipper and W2 domains 1
Gene Aliases
BZAP45, Nbla10236
UniGene
Hs.355983
Gene Chromosome Location
Chr.2: 200811546-200823851 on Build GRCh38
Transcripts
NM_001207067.1 NM_001207069.1 NM_001321688.1 NM_001321690.1 NM_001321691.1 NM_001321693.1 NM_001321694.1 NM_014670.3 XM_017005393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666466 c.732delT p.D244fs*4 Chr.2: 200818071-200818071 on Build GRCh38
1014748 c.557C>A p.A186D Chr.2: 200817992-200817992 on Build GRCh38
1404419 c.620G>A p.R207Q Chr.2: 200818055-200818055 on Build GRCh38
4666467 c.636delT p.D212fs*4 Chr.2: 200818071-200818071 on Build GRCh38
1591667 c.653C>A p.A218D Chr.2: 200817992-200817992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986107458 Chr.2: 200817958-200817958 on Build GRCh38
rs773025667 Chr.2: 200817968-200817968 on Build GRCh38
rs925180728 Chr.2: 200818089-200818089 on Build GRCh38
rs764251679 Chr.2: 200818093-200818093 on Build GRCh38
rs956974608 Chr.2: 200817895-200817895 on Build GRCh38
rs765144282 Chr.2: 200818026-200818026 on Build GRCh38
rs371155481 Chr.2: 200817970-200817970 on Build GRCh38
rs770487405 Chr.2: 200817971-200817971 on Build GRCh38
rs752459717 Chr.2: 200818046-200818046 on Build GRCh38
rs751794822 Chr.2: 200818100-200818100 on Build GRCh38
rs1050420087 Chr.2: 200818095-200818095 on Build GRCh38
rs868599904 Chr.2: 200818037-200818037 on Build GRCh38
rs960456246 Chr.2: 200817910-200817910 on Build GRCh38
rs915497554 Chr.2: 200817918-200817918 on Build GRCh38
rs990105145 Chr.2: 200817906-200817906 on Build GRCh38
rs749716700 Chr.2: 200817943-200817943 on Build GRCh38
rs759511874 Chr.2: 200818011-200818011 on Build GRCh38
rs951715793 Chr.2: 200817890-200817890 on Build GRCh38
rs376417934 Chr.2: 200817954-200817954 on Build GRCh38
rs373755292 Chr.2: 200817937-200817937 on Build GRCh38
rs374224873 Chr.2: 200817999-200817999 on Build GRCh38
rs762768963 Chr.2: 200818078-200818078 on Build GRCh38
rs992405499 Chr.2: 200817915-200817915 on Build GRCh38
rs775085728 Chr.2: 200817939-200817939 on Build GRCh38
rs562499498 Chr.2: 200817914-200817914 on Build GRCh38
rs764044949 Chr.2: 200818082-200818085 on Build GRCh38
rs980409936 Chr.2: 200817892-200817892 on Build GRCh38

View Full Product Details