Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13657466-13657737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
UniGene
Hs.330463
Gene Chromosome Location
Chr.10: 13643706-14330952 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916568 c.2104G>A p.A702T Chr.10: 13657485-13657485 on Build GRCh38
6666915 c.2069C>T p.S690L Chr.10: 13657520-13657520 on Build GRCh38
4685627 c.2089C>T p.R697* Chr.10: 13657500-13657500 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749385274 Chr.10: 13657527-13657528 on Build GRCh38
rs1028714092 Chr.10: 13657587-13657587 on Build GRCh38
rs911574231 Chr.10: 13657610-13657610 on Build GRCh38
rs755692249 Chr.10: 13657562-13657562 on Build GRCh38
rs759653874 Chr.10: 13657535-13657535 on Build GRCh38
rs995020348 Chr.10: 13657586-13657586 on Build GRCh38
rs758569966 Chr.10: 13657540-13657540 on Build GRCh38
rs899416518 Chr.10: 13657695-13657695 on Build GRCh38
rs745506001 Chr.10: 13657505-13657505 on Build GRCh38
rs796359927 Chr.10: 13657675-13657675 on Build GRCh38
rs765535414 Chr.10: 13657537-13657537 on Build GRCh38
rs1004896978 Chr.10: 13657687-13657687 on Build GRCh38
rs182767687 Chr.10: 13657707-13657707 on Build GRCh38
rs552239723 Chr.10: 13657702-13657702 on Build GRCh38
rs554844682 Chr.10: 13657519-13657519 on Build GRCh38
rs75171119 Chr.10: 13657596-13657596 on Build GRCh38
rs11258502 Chr.10: 13657698-13657698 on Build GRCh38
rs1036591216 Chr.10: 13657688-13657688 on Build GRCh38
rs757288894 Chr.10: 13657548-13657548 on Build GRCh38
rs189776436 Chr.10: 13657682-13657682 on Build GRCh38
rs935798224 Chr.10: 13657619-13657619 on Build GRCh38
rs774844638 Chr.10: 13657518-13657518 on Build GRCh38
rs752781020 Chr.10: 13657539-13657539 on Build GRCh38
rs745729341 Chr.10: 13657554-13657554 on Build GRCh38
rs772529214 Chr.10: 13657521-13657521 on Build GRCh38
rs568164325 Chr.10: 13657522-13657522 on Build GRCh38
rs1025776786 Chr.10: 13657527-13657527 on Build GRCh38
rs977714364 Chr.10: 13657614-13657614 on Build GRCh38
rs574709382 Chr.10: 13657664-13657664 on Build GRCh38
rs780839712 Chr.10: 13657552-13657552 on Build GRCh38
rs561588845 Chr.10: 13657658-13657658 on Build GRCh38
rs558725322 Chr.10: 13657611-13657611 on Build GRCh38
rs532109944 Chr.10: 13657699-13657699 on Build GRCh38
rs534651878 Chr.10: 13657486-13657486 on Build GRCh38
rs967496713 Chr.10: 13657672-13657672 on Build GRCh38
rs773732567 Chr.10: 13657528-13657528 on Build GRCh38
rs751550552 Chr.10: 13657546-13657546 on Build GRCh38
rs572403817 Chr.10: 13657618-13657618 on Build GRCh38
rs556707964 Chr.10: 13657580-13657580 on Build GRCh38
rs760974383 Chr.10: 13657534-13657534 on Build GRCh38
rs61833193 Chr.10: 13657493-13657493 on Build GRCh38
rs111460439 Chr.10: 13657629-13657629 on Build GRCh38
rs764062166 Chr.10: 13657545-13657545 on Build GRCh38
rs541366545 Chr.10: 13657643-13657643 on Build GRCh38
rs114781925 Chr.10: 13657595-13657595 on Build GRCh38
rs537201829 Chr.10: 13657565-13657565 on Build GRCh38
rs963508713 Chr.10: 13657571-13657571 on Build GRCh38
rs901293411 Chr.10: 13657520-13657520 on Build GRCh38
rs948174650 Chr.10: 13657666-13657666 on Build GRCh38
rs780832289 Chr.10: 13657500-13657500 on Build GRCh38
rs769552010 Chr.10: 13657516-13657516 on Build GRCh38

View Full Product Details