Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21515770-21515995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIDA1
Entrez Gene ID
387640
Gene Name
SKI/DACH domain containing 1
Gene Aliases
C10orf140, DLN-1
UniGene
Hs.350848
Gene Chromosome Location
Chr.10: 21513475-21525682 on Build GRCh38
Transcripts
NM_207371.3 XM_005252450.3 XM_011519480.2 XM_017016225.1 XM_017016226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1627434 c.2052T>C p.N684N Chr.10: 21515771-21515771 on Build GRCh38
917004 c.1953G>A p.S651S Chr.10: 21515870-21515870 on Build GRCh38
4186705 c.2051delA p.N684fs*6 Chr.10: 21515772-21515772 on Build GRCh38
2132819 c.2025C>T p.G675G Chr.10: 21515798-21515798 on Build GRCh38
6310709 c.2047C>A p.H683N Chr.10: 21515776-21515776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749415868 Chr.10: 21515905-21515905 on Build GRCh38
rs781427369 Chr.10: 21515878-21515878 on Build GRCh38
rs764659727 Chr.10: 21515967-21515967 on Build GRCh38
rs376188457 Chr.10: 21515860-21515860 on Build GRCh38
rs768675440 Chr.10: 21515821-21515821 on Build GRCh38
rs1026025706 Chr.10: 21515811-21515811 on Build GRCh38
rs760417503 Chr.10: 21515840-21515840 on Build GRCh38
rs756397303 Chr.10: 21515885-21515885 on Build GRCh38
rs750778030 Chr.10: 21515881-21515881 on Build GRCh38
rs759188612 Chr.10: 21515963-21515963 on Build GRCh38
rs371144284 Chr.10: 21515798-21515798 on Build GRCh38
rs753463632 Chr.10: 21515859-21515859 on Build GRCh38
rs200289456 Chr.10: 21515939-21515939 on Build GRCh38
rs775606721 Chr.10: 21515807-21515807 on Build GRCh38
rs202182877 Chr.10: 21515802-21515802 on Build GRCh38
rs766114403 Chr.10: 21515841-21515841 on Build GRCh38
rs780221882 Chr.10: 21515903-21515903 on Build GRCh38
rs768638933 Chr.10: 21515907-21515907 on Build GRCh38
rs757716590 Chr.10: 21515877-21515877 on Build GRCh38
rs760540380 Chr.10: 21515945-21515945 on Build GRCh38
rs749368720 Chr.10: 21515818-21515818 on Build GRCh38
rs779007202 Chr.10: 21515908-21515908 on Build GRCh38
rs771790989 Chr.10: 21515827-21515827 on Build GRCh38
rs776447632 Chr.10: 21515961-21515961 on Build GRCh38
rs192189211 Chr.10: 21515922-21515922 on Build GRCh38
rs201474324 Chr.10: 21515839-21515839 on Build GRCh38
rs761757773 Chr.10: 21515826-21515826 on Build GRCh38
rs372541967 Chr.10: 21515940-21515940 on Build GRCh38
rs201491848 Chr.10: 21515873-21515873 on Build GRCh38
rs770757641 Chr.10: 21515960-21515960 on Build GRCh38
rs959147065 Chr.10: 21515809-21515809 on Build GRCh38
rs768841628 Chr.10: 21515903-21515905 on Build GRCh38

View Full Product Details