Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139537317-139537554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
UniGene
Hs.522798
Gene Chromosome Location
Chr.X: 139530720-139563459 on Build GRCh38
Transcripts
NM_000133.3 NM_001313913.1 XM_005262397.4

SNP Detail

SNP ID Chromosome Location
rs976283264 Chr.X: 139537405-139537405 on Build GRCh38
rs941329798 Chr.X: 139537529-139537529 on Build GRCh38
rs753662867 Chr.X: 139537456-139537456 on Build GRCh38
rs757175658 Chr.X: 139537488-139537488 on Build GRCh38
rs1047163231 Chr.X: 139537454-139537454 on Build GRCh38
rs1043359261 Chr.X: 139537465-139537465 on Build GRCh38
rs890224467 Chr.X: 139537502-139537502 on Build GRCh38
rs921828129 Chr.X: 139537482-139537482 on Build GRCh38
rs751614353 Chr.X: 139537406-139537406 on Build GRCh38
rs142980421 Chr.X: 139537382-139537382 on Build GRCh38
rs4149680 Chr.X: 139537495-139537495 on Build GRCh38
rs775962002 Chr.X: 139537354-139537354 on Build GRCh38
rs181583009 Chr.X: 139537418-139537418 on Build GRCh38
rs767087007 Chr.X: 139537419-139537419 on Build GRCh38
rs750172813 Chr.X: 139537420-139537420 on Build GRCh38
rs587776735 Chr.X: 139537388-139537388 on Build GRCh38
rs934070817 Chr.X: 139537450-139537450 on Build GRCh38
rs201895544 Chr.X: 139537430-139537430 on Build GRCh38
rs186259903 Chr.X: 139537512-139537512 on Build GRCh38
rs988784728 Chr.X: 139537385-139537385 on Build GRCh38
rs764166138 Chr.X: 139537440-139537440 on Build GRCh38
rs770212542 Chr.X: 139537350-139537350 on Build GRCh38
rs766328088 Chr.X: 139537434-139537434 on Build GRCh38
rs451281 Chr.X: 139537527-139537527 on Build GRCh38
rs1004613362 Chr.X: 139537510-139537510 on Build GRCh38

View Full Product Details