Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38337455-38337694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFLAM
Entrez Gene ID
133584
Gene Name
EGF like, fibronectin type III and laminin G domains
Gene Aliases
AGRINL, AGRNL, PIKA
UniGene
Hs.20103
Gene Chromosome Location
Chr.5: 38258409-38465480 on Build GRCh38
Transcripts
NM_001205301.1 NM_152403.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6529480 c.199G>C p.G67R Chr.5: 38337621-38337621 on Build GRCh38
1695644 c.149C>T p.A50V Chr.5: 38337571-38337571 on Build GRCh38
4889335 c.171G>A p.M57I Chr.5: 38337593-38337593 on Build GRCh38
2688747 c.106G>A p.G36R Chr.5: 38337528-38337528 on Build GRCh38
1328803 c.188G>C p.S63T Chr.5: 38337610-38337610 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758313552 Chr.5: 38337486-38337486 on Build GRCh38
rs757348529 Chr.5: 38337500-38337500 on Build GRCh38
rs779458784 Chr.5: 38337622-38337622 on Build GRCh38
rs145509203 Chr.5: 38337669-38337669 on Build GRCh38
rs182712141 Chr.5: 38337525-38337525 on Build GRCh38
rs754196781 Chr.5: 38337598-38337598 on Build GRCh38
rs76403205 Chr.5: 38337511-38337511 on Build GRCh38
rs763958349 Chr.5: 38337488-38337488 on Build GRCh38
rs376475358 Chr.5: 38337516-38337516 on Build GRCh38
rs746520691 Chr.5: 38337546-38337546 on Build GRCh38
rs373441112 Chr.5: 38337640-38337640 on Build GRCh38
rs149228822 Chr.5: 38337575-38337575 on Build GRCh38
rs751333691 Chr.5: 38337548-38337552 on Build GRCh38
rs772637164 Chr.5: 38337639-38337639 on Build GRCh38
rs756282751 Chr.5: 38337583-38337583 on Build GRCh38
rs746032022 Chr.5: 38337507-38337507 on Build GRCh38
rs774903237 Chr.5: 38337592-38337592 on Build GRCh38
rs779326065 Chr.5: 38337536-38337536 on Build GRCh38
rs762797656 Chr.5: 38337658-38337658 on Build GRCh38
rs780305435 Chr.5: 38337589-38337589 on Build GRCh38
rs750537300 Chr.5: 38337581-38337581 on Build GRCh38
rs768279215 Chr.5: 38337662-38337662 on Build GRCh38
rs751509580 Chr.5: 38337569-38337569 on Build GRCh38
rs759197022 Chr.5: 38337554-38337554 on Build GRCh38
rs373514546 Chr.5: 38337642-38337642 on Build GRCh38
rs769186445 Chr.5: 38337529-38337529 on Build GRCh38
rs925640657 Chr.5: 38337520-38337520 on Build GRCh38
rs985671065 Chr.5: 38337512-38337512 on Build GRCh38
rs376594419 Chr.5: 38337552-38337552 on Build GRCh38
rs775352358 Chr.5: 38337657-38337657 on Build GRCh38
rs775374937 Chr.5: 38337565-38337565 on Build GRCh38
rs887241792 Chr.5: 38337494-38337494 on Build GRCh38
rs748541066 Chr.5: 38337632-38337632 on Build GRCh38
rs145406817 Chr.5: 38337568-38337568 on Build GRCh38
rs755361364 Chr.5: 38337613-38337613 on Build GRCh38
rs766301604 Chr.5: 38337487-38337488 on Build GRCh38
rs115153941 Chr.5: 38337650-38337650 on Build GRCh38
rs774201319 Chr.5: 38337666-38337666 on Build GRCh38
rs149682724 Chr.5: 38337495-38337495 on Build GRCh38
rs369516889 Chr.5: 38337553-38337553 on Build GRCh38
rs781379853 Chr.5: 38337501-38337501 on Build GRCh38

View Full Product Details