Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114570593-114570860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS2
Entrez Gene ID
10286
Gene Name
breast carcinoma amplified sequence 2
Gene Aliases
DAM1, SPF27, Snt309
UniGene
Hs.22960
Gene Chromosome Location
Chr.1: 114567557-114581644 on Build GRCh38
Transcripts
NM_005872.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894194 c.444C>T p.H148H Chr.1: 114570726-114570726 on Build GRCh38
290631 c.470G>A p.R157K Chr.1: 114570700-114570700 on Build GRCh38
462672 c.440A>G p.E147G Chr.1: 114570730-114570730 on Build GRCh38
1472438 c.439G>C p.E147Q Chr.1: 114570731-114570731 on Build GRCh38
7425290 c.439G>A p.E147K Chr.1: 114570731-114570731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957410970 Chr.1: 114570633-114570633 on Build GRCh38
rs954358379 Chr.1: 114570646-114570646 on Build GRCh38
rs777561805 Chr.1: 114570617-114570617 on Build GRCh38
rs200831848 Chr.1: 114570771-114570771 on Build GRCh38
rs904879155 Chr.1: 114570638-114570638 on Build GRCh38
rs761844764 Chr.1: 114570717-114570717 on Build GRCh38
rs780272103 Chr.1: 114570792-114570792 on Build GRCh38
rs372979252 Chr.1: 114570663-114570663 on Build GRCh38
rs368828386 Chr.1: 114570755-114570755 on Build GRCh38
rs757776095 Chr.1: 114570758-114570758 on Build GRCh38
rs527644009 Chr.1: 114570654-114570654 on Build GRCh38
rs990172702 Chr.1: 114570634-114570634 on Build GRCh38
rs767359809 Chr.1: 114570725-114570725 on Build GRCh38
rs1031777392 Chr.1: 114570624-114570627 on Build GRCh38
rs374723628 Chr.1: 114570753-114570753 on Build GRCh38
rs1002253186 Chr.1: 114570645-114570645 on Build GRCh38
rs548954718 Chr.1: 114570748-114570748 on Build GRCh38
rs972848138 Chr.1: 114570814-114570814 on Build GRCh38
rs753725924 Chr.1: 114570726-114570726 on Build GRCh38
rs753268271 Chr.1: 114570740-114570740 on Build GRCh38
rs781524905 Chr.1: 114570762-114570762 on Build GRCh38
rs774302766 Chr.1: 114570710-114570710 on Build GRCh38
rs191689978 Chr.1: 114570637-114570637 on Build GRCh38
rs567368093 Chr.1: 114570776-114570776 on Build GRCh38
rs199743368 Chr.1: 114570728-114570728 on Build GRCh38
rs746271439 Chr.1: 114570784-114570784 on Build GRCh38
rs756307009 Chr.1: 114570791-114570791 on Build GRCh38
rs766100774 Chr.1: 114570737-114570737 on Build GRCh38

View Full Product Details