Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126990428-126990679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239629 c.470A>T p.N157I Chr.10: 126990600-126990600 on Build GRCh38
4839958 c.438G>T p.K146N Chr.10: 126990568-126990568 on Build GRCh38
4678935 c.420T>G p.D140E Chr.10: 126990550-126990550 on Build GRCh38
5468872 c.461A>G p.D154G Chr.10: 126990591-126990591 on Build GRCh38
3985494 c.397C>G p.L133V Chr.10: 126990527-126990527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374693322 Chr.10: 126990518-126990518 on Build GRCh38
rs762939076 Chr.10: 126990623-126990623 on Build GRCh38
rs750631429 Chr.10: 126990498-126990498 on Build GRCh38
rs760460729 Chr.10: 126990475-126990475 on Build GRCh38
rs757300109 Chr.10: 126990486-126990486 on Build GRCh38
rs193160639 Chr.10: 126990477-126990477 on Build GRCh38
rs754046061 Chr.10: 126990640-126990640 on Build GRCh38
rs145105617 Chr.10: 126990593-126990593 on Build GRCh38
rs748731456 Chr.10: 126990560-126990560 on Build GRCh38
rs779529803 Chr.10: 126990530-126990530 on Build GRCh38
rs770644938 Chr.10: 126990588-126990588 on Build GRCh38
rs75555086 Chr.10: 126990558-126990558 on Build GRCh38
rs762258153 Chr.10: 126990623-126990630 on Build GRCh38
rs576434465 Chr.10: 126990570-126990570 on Build GRCh38
rs182891892 Chr.10: 126990630-126990630 on Build GRCh38
rs188061145 Chr.10: 126990632-126990632 on Build GRCh38
rs771234802 Chr.10: 126990507-126990507 on Build GRCh38
rs746572428 Chr.10: 126990585-126990585 on Build GRCh38
rs759349034 Chr.10: 126990483-126990483 on Build GRCh38
rs772859229 Chr.10: 126990581-126990581 on Build GRCh38
rs758997336 Chr.10: 126990564-126990565 on Build GRCh38
rs1048901548 Chr.10: 126990595-126990595 on Build GRCh38
rs772994764 Chr.10: 126990472-126990472 on Build GRCh38
rs950569150 Chr.10: 126990565-126990567 on Build GRCh38
rs755364312 Chr.10: 126990646-126990646 on Build GRCh38
rs759356702 Chr.10: 126990596-126990596 on Build GRCh38
rs766026803 Chr.10: 126990476-126990476 on Build GRCh38
rs780941985 Chr.10: 126990515-126990515 on Build GRCh38
rs537707948 Chr.10: 126990601-126990601 on Build GRCh38
rs773583444 Chr.10: 126990523-126990523 on Build GRCh38
rs779478716 Chr.10: 126990650-126990650 on Build GRCh38
rs766647410 Chr.10: 126990639-126990639 on Build GRCh38
rs749695334 Chr.10: 126990516-126990516 on Build GRCh38
rs764984706 Chr.10: 126990597-126990597 on Build GRCh38
rs192503473 Chr.10: 126990634-126990634 on Build GRCh38
rs765245181 Chr.10: 126990485-126990485 on Build GRCh38
rs973298505 Chr.10: 126990482-126990482 on Build GRCh38
rs199534710 Chr.10: 126990631-126990631 on Build GRCh38
rs767654005 Chr.10: 126990487-126990487 on Build GRCh38

View Full Product Details