Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100153441-100153686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMTOR4
Entrez Gene ID
389541
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
Gene Aliases
C7orf59
UniGene
Hs.406520
Gene Chromosome Location
Chr.7: 100130969-100154212 on Build GRCh38
Transcripts
NM_001008395.3 NM_001318236.1 NM_001318237.1 NR_134526.1 NR_134527.1 NR_134528.1 NR_134529.1 NR_134530.1 XM_017012198.1 XM_017012199.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5976549 c.171C>T p.R57R Chr.7: 100153486-100153486 on Build GRCh38
374586 c.182T>C p.V61A Chr.7: 100153497-100153497 on Build GRCh38
5429665 c.200C>G p.S67C Chr.7: 100153515-100153515 on Build GRCh38
7276631 c.154G>A p.G52S Chr.7: 100153469-100153469 on Build GRCh38
6699501 c.160C>T p.R54W Chr.7: 100153475-100153475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766916913 Chr.7: 100153556-100153556 on Build GRCh38
rs763235564 Chr.7: 100153625-100153625 on Build GRCh38
rs575298780 Chr.7: 100153611-100153611 on Build GRCh38
rs756025539 Chr.7: 100153539-100153539 on Build GRCh38
rs750435063 Chr.7: 100153469-100153469 on Build GRCh38
rs768047458 Chr.7: 100153575-100153575 on Build GRCh38
rs956174312 Chr.7: 100153644-100153644 on Build GRCh38
rs145738939 Chr.7: 100153468-100153468 on Build GRCh38
rs747870612 Chr.7: 100153484-100153484 on Build GRCh38
rs150613556 Chr.7: 100153617-100153617 on Build GRCh38
rs767646400 Chr.7: 100153521-100153522 on Build GRCh38
rs781688722 Chr.7: 100153481-100153481 on Build GRCh38
rs752622555 Chr.7: 100153575-100153582 on Build GRCh38
rs770925024 Chr.7: 100153500-100153500 on Build GRCh38
rs771912815 Chr.7: 100153525-100153525 on Build GRCh38
rs762058054 Chr.7: 100153531-100153531 on Build GRCh38
rs773088960 Chr.7: 100153482-100153482 on Build GRCh38
rs556679538 Chr.7: 100153606-100153606 on Build GRCh38
rs777244135 Chr.7: 100153583-100153583 on Build GRCh38
rs759141796 Chr.7: 100153582-100153582 on Build GRCh38
rs755329254 Chr.7: 100153480-100153480 on Build GRCh38
rs771966105 Chr.7: 100153607-100153607 on Build GRCh38
rs774087460 Chr.7: 100153519-100153519 on Build GRCh38
rs775244079 Chr.7: 100153526-100153526 on Build GRCh38
rs758972020 Chr.7: 100153470-100153470 on Build GRCh38
rs755807219 Chr.7: 100153485-100153485 on Build GRCh38
rs368360114 Chr.7: 100153465-100153465 on Build GRCh38
rs924603286 Chr.7: 100153471-100153471 on Build GRCh38
rs200068622 Chr.7: 100153547-100153547 on Build GRCh38
rs766744606 Chr.7: 100153475-100153475 on Build GRCh38
rs750534413 Chr.7: 100153556-100153557 on Build GRCh38
rs35195309 Chr.7: 100153576-100153576 on Build GRCh38
rs777383874 Chr.7: 100153487-100153487 on Build GRCh38
rs921814362 Chr.7: 100153520-100153520 on Build GRCh38
rs373505176 Chr.7: 100153557-100153557 on Build GRCh38
rs76212539 Chr.7: 100153656-100153656 on Build GRCh38
rs753079902 Chr.7: 100153577-100153577 on Build GRCh38
rs200961392 Chr.7: 100153491-100153491 on Build GRCh38
rs3736590 Chr.7: 100153658-100153658 on Build GRCh38
rs765203988 Chr.7: 100153558-100153558 on Build GRCh38
rs201730556 Chr.7: 100153558-100153558 on Build GRCh38
rs770977405 Chr.7: 100153637-100153637 on Build GRCh38
rs200395313 Chr.7: 100153476-100153476 on Build GRCh38
rs534619483 Chr.7: 100153524-100153524 on Build GRCh38
rs386716113 Chr.7: 100153656-100153658 on Build GRCh38
rs756782291 Chr.7: 100153594-100153594 on Build GRCh38
rs1048630684 Chr.7: 100153511-100153511 on Build GRCh38
rs935113932 Chr.7: 100153571-100153571 on Build GRCh38
rs753403678 Chr.7: 100153588-100153588 on Build GRCh38
rs778471636 Chr.7: 100153599-100153599 on Build GRCh38
rs760110999 Chr.7: 100153563-100153563 on Build GRCh38
rs552877824 Chr.7: 100153535-100153535 on Build GRCh38
rs775162710 Chr.7: 100153618-100153618 on Build GRCh38
rs756325888 Chr.7: 100153557-100153558 on Build GRCh38

View Full Product Details