Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135574535-135574774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL14
Entrez Gene ID
9547
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14
UniGene
Hs.483444
Gene Chromosome Location
Chr.5: 135570679-135579279 on Build GRCh38
Transcripts
NM_004887.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7273869 c.301G>A p.A101T Chr.5: 135574591-135574591 on Build GRCh38
1310560 c.309C>T p.N103N Chr.5: 135574583-135574583 on Build GRCh38
7327800 c.320+9C>T p.? Chr.5: 135574563-135574563 on Build GRCh38
7213534 c.230G>A p.R77K Chr.5: 135574662-135574662 on Build GRCh38
4616323 c.236G>A p.R79Q Chr.5: 135574656-135574656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976783688 Chr.5: 135574574-135574574 on Build GRCh38
rs781182895 Chr.5: 135574642-135574642 on Build GRCh38
rs903451361 Chr.5: 135574753-135574753 on Build GRCh38
rs146115267 Chr.5: 135574674-135574674 on Build GRCh38
rs151075257 Chr.5: 135574573-135574573 on Build GRCh38
rs535801773 Chr.5: 135574719-135574719 on Build GRCh38
rs755238282 Chr.5: 135574589-135574589 on Build GRCh38
rs769164567 Chr.5: 135574554-135574554 on Build GRCh38
rs765232019 Chr.5: 135574591-135574591 on Build GRCh38
rs150851659 Chr.5: 135574592-135574592 on Build GRCh38
rs769556145 Chr.5: 135574656-135574656 on Build GRCh38
rs765446726 Chr.5: 135574685-135574685 on Build GRCh38
rs988209044 Chr.5: 135574567-135574567 on Build GRCh38
rs779706762 Chr.5: 135574718-135574718 on Build GRCh38
rs757089753 Chr.5: 135574627-135574627 on Build GRCh38
rs751522250 Chr.5: 135574697-135574697 on Build GRCh38
rs762222233 Chr.5: 135574563-135574563 on Build GRCh38
rs372169387 Chr.5: 135574691-135574691 on Build GRCh38
rs747497914 Chr.5: 135574733-135574733 on Build GRCh38
rs865877942 Chr.5: 135574586-135574586 on Build GRCh38
rs985924883 Chr.5: 135574566-135574566 on Build GRCh38
rs768072701 Chr.5: 135574660-135574660 on Build GRCh38
rs768178618 Chr.5: 135574721-135574721 on Build GRCh38
rs745502995 Chr.5: 135574654-135574654 on Build GRCh38
rs750176422 Chr.5: 135574712-135574712 on Build GRCh38
rs778439853 Chr.5: 135574732-135574732 on Build GRCh38
rs766758665 Chr.5: 135574580-135574580 on Build GRCh38
rs373520365 Chr.5: 135574713-135574713 on Build GRCh38
rs748772776 Chr.5: 135574657-135574657 on Build GRCh38
rs371288714 Chr.5: 135574700-135574700 on Build GRCh38
rs115896115 Chr.5: 135574568-135574568 on Build GRCh38
rs747049288 Chr.5: 135574562-135574562 on Build GRCh38
rs17168623 Chr.5: 135574692-135574692 on Build GRCh38
rs773665537 Chr.5: 135574661-135574661 on Build GRCh38
rs546943314 Chr.5: 135574669-135574669 on Build GRCh38
rs1032655009 Chr.5: 135574558-135574558 on Build GRCh38
rs200916552 Chr.5: 135574575-135574575 on Build GRCh38
rs139612389 Chr.5: 135574611-135574611 on Build GRCh38
rs781420160 Chr.5: 135574594-135574594 on Build GRCh38
rs369589670 Chr.5: 135574702-135574702 on Build GRCh38
rs754024461 Chr.5: 135574582-135574582 on Build GRCh38
rs751579907 Chr.5: 135574616-135574616 on Build GRCh38
rs764122117 Chr.5: 135574693-135574693 on Build GRCh38
rs761207832 Chr.5: 135574667-135574667 on Build GRCh38
rs759827097 Chr.5: 135574681-135574681 on Build GRCh38

View Full Product Details