Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31933698-31933965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NM_001282459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624864 c.698C>T p.P233L Chr.6: 31933948-31933948 on Build GRCh38
6615493 c.600G>A p.T200T Chr.6: 31933767-31933767 on Build GRCh38
4504830 c.675C>T p.S225S Chr.6: 31933925-31933925 on Build GRCh38
7358359 c.553G>T p.G185W Chr.6: 31933720-31933720 on Build GRCh38
4006373 c.645C>T p.D215D Chr.6: 31933895-31933895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751704022 Chr.6: 31933816-31933816 on Build GRCh38
rs777520299 Chr.6: 31933860-31933860 on Build GRCh38
rs764340299 Chr.6: 31933733-31933733 on Build GRCh38
rs769177291 Chr.6: 31933922-31933922 on Build GRCh38
rs202082500 Chr.6: 31933759-31933759 on Build GRCh38
rs754916117 Chr.6: 31933830-31933830 on Build GRCh38
rs759460529 Chr.6: 31933750-31933750 on Build GRCh38
rs771778366 Chr.6: 31933761-31933761 on Build GRCh38
rs201851644 Chr.6: 31933826-31933826 on Build GRCh38
rs762058437 Chr.6: 31933821-31933821 on Build GRCh38
rs758166636 Chr.6: 31933847-31933847 on Build GRCh38
rs774870557 Chr.6: 31933744-31933744 on Build GRCh38
rs751723882 Chr.6: 31933740-31933740 on Build GRCh38
rs778818261 Chr.6: 31933835-31933835 on Build GRCh38
rs760731117 Chr.6: 31933937-31933937 on Build GRCh38
rs764464539 Chr.6: 31933813-31933813 on Build GRCh38
rs752545777 Chr.6: 31933838-31933838 on Build GRCh38
rs1008619459 Chr.6: 31933833-31933833 on Build GRCh38
rs141450429 Chr.6: 31933914-31933914 on Build GRCh38
rs770315100 Chr.6: 31933780-31933780 on Build GRCh38
rs770363423 Chr.6: 31933796-31933796 on Build GRCh38
rs147186833 Chr.6: 31933781-31933781 on Build GRCh38
rs761956730 Chr.6: 31933926-31933926 on Build GRCh38
rs745306334 Chr.6: 31933906-31933906 on Build GRCh38
rs775998443 Chr.6: 31933802-31933802 on Build GRCh38
rs750536670 Chr.6: 31933936-31933936 on Build GRCh38
rs753059312 Chr.6: 31933727-31933727 on Build GRCh38
rs778695362 Chr.6: 31933753-31933753 on Build GRCh38
rs770485901 Chr.6: 31933911-31933911 on Build GRCh38
rs28934590 Chr.6: 31933876-31933876 on Build GRCh38
rs1004323059 Chr.6: 31933890-31933890 on Build GRCh38
rs945580226 Chr.6: 31933784-31933784 on Build GRCh38
rs757266578 Chr.6: 31933746-31933746 on Build GRCh38
rs900164037 Chr.6: 31933868-31933868 on Build GRCh38
rs774928519 Chr.6: 31933837-31933837 on Build GRCh38
rs771542146 Chr.6: 31933822-31933822 on Build GRCh38
rs772837862 Chr.6: 31933766-31933766 on Build GRCh38
rs148330562 Chr.6: 31933887-31933887 on Build GRCh38
rs549362270 Chr.6: 31933938-31933938 on Build GRCh38
rs759980745 Chr.6: 31933783-31933783 on Build GRCh38
rs763454963 Chr.6: 31933915-31933915 on Build GRCh38
rs996799187 Chr.6: 31933855-31933855 on Build GRCh38
rs199946097 Chr.6: 31933732-31933732 on Build GRCh38
rs767714124 Chr.6: 31933931-31933931 on Build GRCh38
rs759972430 Chr.6: 31933806-31933806 on Build GRCh38
rs750461415 Chr.6: 31933825-31933825 on Build GRCh38
rs774731673 Chr.6: 31933923-31933923 on Build GRCh38
rs749217491 Chr.6: 31933749-31933749 on Build GRCh38
rs372423916 Chr.6: 31933880-31933880 on Build GRCh38
rs903329492 Chr.6: 31933785-31933785 on Build GRCh38
rs765371189 Chr.6: 31933923-31933924 on Build GRCh38
rs754863047 Chr.6: 31933751-31933751 on Build GRCh38
rs949860132 Chr.6: 31933770-31933770 on Build GRCh38
rs746577678 Chr.6: 31933776-31933776 on Build GRCh38

View Full Product Details