Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14020153-14020386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN8
Entrez Gene ID
54960
Gene Name
gem nuclear organelle associated protein 8
Gene Aliases
FAM51A1
UniGene
Hs.592237
Gene Chromosome Location
Chr.X: 14002494-14030087 on Build GRCh38
Transcripts
NM_001042479.1 NM_001042480.1 NM_017856.2 XM_005274555.2 XM_017029617.1 XM_017029618.1

SNP Detail

SNP ID Chromosome Location
rs757345566 Chr.X: 14020297-14020297 on Build GRCh38
rs765879777 Chr.X: 14020248-14020248 on Build GRCh38
rs779033647 Chr.X: 14020314-14020314 on Build GRCh38
rs747399327 Chr.X: 14020338-14020338 on Build GRCh38
rs769319019 Chr.X: 14020239-14020239 on Build GRCh38
rs893616500 Chr.X: 14020183-14020183 on Build GRCh38
rs200576997 Chr.X: 14020263-14020263 on Build GRCh38
rs747311447 Chr.X: 14020186-14020186 on Build GRCh38
rs947384749 Chr.X: 14020361-14020361 on Build GRCh38
rs151153824 Chr.X: 14020249-14020249 on Build GRCh38
rs752648197 Chr.X: 14020276-14020276 on Build GRCh38
rs145346660 Chr.X: 14020337-14020337 on Build GRCh38
rs367998555 Chr.X: 14020346-14020346 on Build GRCh38
rs762654716 Chr.X: 14020242-14020242 on Build GRCh38
rs371906124 Chr.X: 14020351-14020351 on Build GRCh38
rs769187670 Chr.X: 14020189-14020189 on Build GRCh38
rs1020656249 Chr.X: 14020247-14020247 on Build GRCh38
rs772807636 Chr.X: 14020240-14020240 on Build GRCh38
rs137990246 Chr.X: 14020358-14020358 on Build GRCh38
rs140266909 Chr.X: 14020328-14020328 on Build GRCh38
rs1018585197 Chr.X: 14020298-14020298 on Build GRCh38
rs748686919 Chr.X: 14020352-14020352 on Build GRCh38
rs761363023 Chr.X: 14020216-14020216 on Build GRCh38
rs767611226 Chr.X: 14020270-14020270 on Build GRCh38
rs143025002 Chr.X: 14020200-14020200 on Build GRCh38
rs750408682 Chr.X: 14020321-14020321 on Build GRCh38

View Full Product Details