Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109556776-109557009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBAG9
Entrez Gene ID
9166
Gene Name
estrogen receptor binding site associated, antigen, 9
Gene Aliases
EB9, PDAF
UniGene
Hs.409368
Gene Chromosome Location
Chr.8: 109539700-109565996 on Build GRCh38
Transcripts
NM_001278938.1 NM_004215.4 NM_198120.2 XM_017013960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4544984 c.367G>A p.G123R Chr.8: 109556980-109556980 on Build GRCh38
1094985 c.340G>T p.E114* Chr.8: 109556953-109556953 on Build GRCh38
461497 c.364G>C p.D122H Chr.8: 109556977-109556977 on Build GRCh38
3884726 c.348G>T p.L116F Chr.8: 109556961-109556961 on Build GRCh38
4680003 c.384T>C p.S128S Chr.8: 109556997-109556997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756671301 Chr.8: 109556912-109556912 on Build GRCh38
rs781683869 Chr.8: 109556903-109556903 on Build GRCh38
rs182265187 Chr.8: 109556895-109556895 on Build GRCh38
rs201700353 Chr.8: 109556921-109556921 on Build GRCh38
rs372276656 Chr.8: 109556979-109556979 on Build GRCh38
rs1044007074 Chr.8: 109556978-109556978 on Build GRCh38
rs751744225 Chr.8: 109556894-109556894 on Build GRCh38
rs945618853 Chr.8: 109556967-109556967 on Build GRCh38
rs550433240 Chr.8: 109556804-109556804 on Build GRCh38
rs758704614 Chr.8: 109556937-109556937 on Build GRCh38
rs985849961 Chr.8: 109556858-109556858 on Build GRCh38
rs563652595 Chr.8: 109556870-109556870 on Build GRCh38
rs749793004 Chr.8: 109556959-109556959 on Build GRCh38
rs780564381 Chr.8: 109556954-109556954 on Build GRCh38
rs569999297 Chr.8: 109556877-109556877 on Build GRCh38
rs951715525 Chr.8: 109556825-109556825 on Build GRCh38
rs778083347 Chr.8: 109556817-109556820 on Build GRCh38
rs750861557 Chr.8: 109556908-109556908 on Build GRCh38
rs866321465 Chr.8: 109556974-109556974 on Build GRCh38
rs914110273 Chr.8: 109556942-109556942 on Build GRCh38
rs994256722 Chr.8: 109556916-109556916 on Build GRCh38
rs757607217 Chr.8: 109556897-109556897 on Build GRCh38

View Full Product Details