Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224770206-224770475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
UniGene
Hs.46578
Gene Chromosome Location
Chr.2: 224765090-225042689 on Build GRCh38
Transcripts
NM_001290263.1 NM_014689.2 XM_005246673.4 XM_006712617.3 XM_017004441.1 XM_017004442.1 XM_017004443.1 XM_017004444.1 XM_017004445.1 XM_017004446.1 XM_017004447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6467018 c.6387G>A p.R2129R Chr.2: 224770262-224770262 on Build GRCh38
6157295 c.1849C>A p.Q617K Chr.2: 224770327-224770327 on Build GRCh38
5948655 c.6370C>T p.L2124L Chr.2: 224770267-224770267 on Build GRCh38
4807028 c.6351C>G p.I2117M Chr.2: 224770298-224770298 on Build GRCh38
294324 c.6391C>T p.H2131Y Chr.2: 224770258-224770258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747128792 Chr.2: 224770393-224770393 on Build GRCh38
rs755926775 Chr.2: 224770340-224770340 on Build GRCh38
rs766124100 Chr.2: 224770357-224770357 on Build GRCh38
rs1043279022 Chr.2: 224770443-224770443 on Build GRCh38
rs770230430 Chr.2: 224770314-224770314 on Build GRCh38
rs938305508 Chr.2: 224770370-224770370 on Build GRCh38
rs138819761 Chr.2: 224770316-224770316 on Build GRCh38
rs375933534 Chr.2: 224770346-224770346 on Build GRCh38
rs547710815 Chr.2: 224770315-224770315 on Build GRCh38
rs558501585 Chr.2: 224770404-224770404 on Build GRCh38
rs750056919 Chr.2: 224770338-224770338 on Build GRCh38
rs569974511 Chr.2: 224770388-224770388 on Build GRCh38
rs780016487 Chr.2: 224770253-224770253 on Build GRCh38
rs772172610 Chr.2: 224770262-224770262 on Build GRCh38
rs749175174 Chr.2: 224770256-224770256 on Build GRCh38
rs1031799769 Chr.2: 224770356-224770356 on Build GRCh38
rs768886621 Chr.2: 224770321-224770321 on Build GRCh38
rs145037309 Chr.2: 224770247-224770247 on Build GRCh38
rs889592816 Chr.2: 224770448-224770448 on Build GRCh38
rs771118606 Chr.2: 224770305-224770305 on Build GRCh38
rs200577403 Chr.2: 224770237-224770237 on Build GRCh38
rs751226569 Chr.2: 224770235-224770235 on Build GRCh38
rs777648661 Chr.2: 224770281-224770281 on Build GRCh38
rs369381199 Chr.2: 224770309-224770309 on Build GRCh38
rs780883039 Chr.2: 224770238-224770238 on Build GRCh38
rs773789129 Chr.2: 224770322-224770322 on Build GRCh38
rs776912091 Chr.2: 224770306-224770306 on Build GRCh38
rs76798202 Chr.2: 224770406-224770406 on Build GRCh38
rs947192345 Chr.2: 224770429-224770429 on Build GRCh38
rs16866164 Chr.2: 224770389-224770389 on Build GRCh38
rs536592870 Chr.2: 224770330-224770330 on Build GRCh38
rs550441284 Chr.2: 224770336-224770336 on Build GRCh38
rs747271237 Chr.2: 224770286-224770286 on Build GRCh38
rs199583328 Chr.2: 224770323-224770323 on Build GRCh38
rs775881219 Chr.2: 224770414-224770414 on Build GRCh38
rs753781556 Chr.2: 224770364-224770364 on Build GRCh38
rs984685244 Chr.2: 224770304-224770304 on Build GRCh38

View Full Product Details