Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10983772-10984011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL2
Entrez Gene ID
54898
Gene Name
ELOVL fatty acid elongase 2
Gene Aliases
SSC2
UniGene
Hs.656436
Gene Chromosome Location
Chr.6: 10980759-11044391 on Build GRCh38
Transcripts
NM_017770.3 XM_011514716.2 XM_011514717.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1072019 c.766-1G>T p.? Chr.6: 10983907-10983907 on Build GRCh38
6172199 c.832G>A p.G278S Chr.6: 10983840-10983840 on Build GRCh38
1439747 c.856G>A p.A286T Chr.6: 10983816-10983816 on Build GRCh38
1439748 c.773G>A p.R258Q Chr.6: 10983899-10983899 on Build GRCh38
1439749 c.772C>T p.R258* Chr.6: 10983900-10983900 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774747090 Chr.6: 10983899-10983899 on Build GRCh38
rs143359371 Chr.6: 10983875-10983875 on Build GRCh38
rs762218875 Chr.6: 10983903-10983903 on Build GRCh38
rs376180925 Chr.6: 10983915-10983915 on Build GRCh38
rs766850068 Chr.6: 10983932-10983932 on Build GRCh38
rs780169856 Chr.6: 10983879-10983879 on Build GRCh38
rs750987217 Chr.6: 10983814-10983814 on Build GRCh38
rs758181725 Chr.6: 10983844-10983848 on Build GRCh38
rs149258164 Chr.6: 10983868-10983868 on Build GRCh38
rs759133129 Chr.6: 10983819-10983819 on Build GRCh38
rs757069139 Chr.6: 10983866-10983866 on Build GRCh38
rs112015080 Chr.6: 10983880-10983880 on Build GRCh38
rs758051642 Chr.6: 10983859-10983859 on Build GRCh38
rs751193491 Chr.6: 10983956-10983956 on Build GRCh38
rs930008592 Chr.6: 10983960-10983960 on Build GRCh38
rs761135020 Chr.6: 10983831-10983831 on Build GRCh38
rs764772741 Chr.6: 10983832-10983832 on Build GRCh38
rs773882982 Chr.6: 10983811-10983811 on Build GRCh38
rs777473383 Chr.6: 10983945-10983946 on Build GRCh38
rs191521211 Chr.6: 10983900-10983900 on Build GRCh38
rs369205080 Chr.6: 10983953-10983953 on Build GRCh38
rs768096314 Chr.6: 10983805-10983805 on Build GRCh38
rs367685105 Chr.6: 10983888-10983888 on Build GRCh38
rs769690398 Chr.6: 10983877-10983877 on Build GRCh38
rs777413957 Chr.6: 10983860-10983860 on Build GRCh38
rs762371530 Chr.6: 10983939-10983939 on Build GRCh38
rs550159915 Chr.6: 10983974-10983974 on Build GRCh38
rs775003533 Chr.6: 10983934-10983934 on Build GRCh38
rs909109838 Chr.6: 10983857-10983857 on Build GRCh38
rs746816044 Chr.6: 10983946-10983949 on Build GRCh38
rs773490276 Chr.6: 10983910-10983910 on Build GRCh38
rs578237717 Chr.6: 10983808-10983808 on Build GRCh38
rs376686946 Chr.6: 10983804-10983804 on Build GRCh38
rs772540713 Chr.6: 10983909-10983909 on Build GRCh38
rs761238962 Chr.6: 10983916-10983916 on Build GRCh38
rs751285613 Chr.6: 10983864-10983864 on Build GRCh38
rs752211623 Chr.6: 10983849-10983849 on Build GRCh38

View Full Product Details