Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10140962-10141219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
UniGene
Hs.202672
Gene Chromosome Location
Chr.19: 10133344-10195135 on Build GRCh38
Transcripts
NM_001130823.2 NM_001318730.1 NM_001318731.1 NM_001379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6974903 c.3262-2A>G p.? Chr.19: 10141191-10141191 on Build GRCh38
6248450 c.3310C>T p.R1104C Chr.19: 10141141-10141141 on Build GRCh38
6936482 c.3345delA p.K1117fs*16 Chr.19: 10141106-10141106 on Build GRCh38
6915736 c.3275A>C p.K1092T Chr.19: 10141176-10141176 on Build GRCh38
4543216 c.3289G>C p.E1097Q Chr.19: 10141162-10141162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989633245 Chr.19: 10140988-10140988 on Build GRCh38
rs750033128 Chr.19: 10141085-10141085 on Build GRCh38
rs756768514 Chr.19: 10141076-10141076 on Build GRCh38
rs1010216337 Chr.19: 10141042-10141042 on Build GRCh38
rs200950656 Chr.19: 10141141-10141141 on Build GRCh38
rs758121423 Chr.19: 10141094-10141094 on Build GRCh38
rs550419332 Chr.19: 10140991-10140991 on Build GRCh38
rs781376510 Chr.19: 10141101-10141101 on Build GRCh38
rs759653796 Chr.19: 10141155-10141155 on Build GRCh38
rs2290684 Chr.19: 10141071-10141071 on Build GRCh38
rs374676749 Chr.19: 10141147-10141147 on Build GRCh38
rs748574658 Chr.19: 10141133-10141133 on Build GRCh38
rs770147435 Chr.19: 10141142-10141142 on Build GRCh38
rs1000261043 Chr.19: 10141098-10141098 on Build GRCh38
rs150331990 Chr.19: 10141146-10141146 on Build GRCh38
rs755865204 Chr.19: 10141156-10141156 on Build GRCh38
rs933543639 Chr.19: 10141128-10141128 on Build GRCh38
rs772372020 Chr.19: 10141194-10141194 on Build GRCh38
rs1021798119 Chr.19: 10140990-10140990 on Build GRCh38
rs967505199 Chr.19: 10141007-10141007 on Build GRCh38
rs1057350237 Chr.19: 10141006-10141006 on Build GRCh38
rs191221466 Chr.19: 10141072-10141072 on Build GRCh38
rs778466510 Chr.19: 10141078-10141078 on Build GRCh38
rs886054128 Chr.19: 10141098-10141098 on Build GRCh38
rs534263445 Chr.19: 10141148-10141148 on Build GRCh38

View Full Product Details