Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70482622-70482883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG4
Entrez Gene ID
25839
Gene Name
component of oligomeric golgi complex 4
Gene Aliases
CDG2J, COD1
UniGene
Hs.208680
Gene Chromosome Location
Chr.16: 70480569-70523554 on Build GRCh38
Transcripts
NM_001195139.1 NM_015386.2 XM_011522981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718451 c.1837_1838AC>TT p.T613L Chr.16: 70482811-70482812 on Build GRCh38
1379309 c.1919_1920+1delAGG p.? Chr.16: 70482728-70482730 on Build GRCh38
273774 c.1910_1912delACA p.N637delN Chr.16: 70482737-70482739 on Build GRCh38
5587516 c.1906C>A p.H636N Chr.16: 70482743-70482743 on Build GRCh38
6891272 c.1828G>A p.E610K Chr.16: 70482821-70482821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370357860 Chr.16: 70482853-70482853 on Build GRCh38
rs931160786 Chr.16: 70482778-70482778 on Build GRCh38
rs766180073 Chr.16: 70482696-70482696 on Build GRCh38
rs758986477 Chr.16: 70482843-70482844 on Build GRCh38
rs1048764460 Chr.16: 70482809-70482809 on Build GRCh38
rs749598979 Chr.16: 70482718-70482718 on Build GRCh38
rs781177606 Chr.16: 70482699-70482699 on Build GRCh38
rs769546927 Chr.16: 70482847-70482847 on Build GRCh38
rs765088543 Chr.16: 70482693-70482693 on Build GRCh38
rs772560552 Chr.16: 70482749-70482749 on Build GRCh38
rs949134176 Chr.16: 70482726-70482726 on Build GRCh38
rs920314679 Chr.16: 70482779-70482779 on Build GRCh38
rs376038606 Chr.16: 70482851-70482851 on Build GRCh38
rs138701123 Chr.16: 70482755-70482755 on Build GRCh38
rs755376017 Chr.16: 70482811-70482811 on Build GRCh38
rs1020463776 Chr.16: 70482654-70482654 on Build GRCh38
rs764121851 Chr.16: 70482798-70482798 on Build GRCh38
rs1041044760 Chr.16: 70482849-70482849 on Build GRCh38
rs796633279 Chr.16: 70482702-70482702 on Build GRCh38
rs74539146 Chr.16: 70482804-70482804 on Build GRCh38
rs780438779 Chr.16: 70482714-70482714 on Build GRCh38
rs565871794 Chr.16: 70482810-70482810 on Build GRCh38
rs371010393 Chr.16: 70482706-70482706 on Build GRCh38
rs900516372 Chr.16: 70482859-70482859 on Build GRCh38
rs754779083 Chr.16: 70482690-70482690 on Build GRCh38
rs746447374 Chr.16: 70482828-70482828 on Build GRCh38
rs377659349 Chr.16: 70482697-70482697 on Build GRCh38
rs756356860 Chr.16: 70482711-70482711 on Build GRCh38
rs554213038 Chr.16: 70482750-70482750 on Build GRCh38
rs771582886 Chr.16: 70482742-70482742 on Build GRCh38
rs748578976 Chr.16: 70482815-70482815 on Build GRCh38
rs750412872 Chr.16: 70482802-70482802 on Build GRCh38
rs201306234 Chr.16: 70482842-70482842 on Build GRCh38
rs758306569 Chr.16: 70482796-70482796 on Build GRCh38
rs140533129 Chr.16: 70482862-70482862 on Build GRCh38
rs770490584 Chr.16: 70482845-70482845 on Build GRCh38
rs776265939 Chr.16: 70482751-70482751 on Build GRCh38
rs778353418 Chr.16: 70482817-70482817 on Build GRCh38
rs968900714 Chr.16: 70482669-70482669 on Build GRCh38
rs373916407 Chr.16: 70482734-70482734 on Build GRCh38
rs1031217880 Chr.16: 70482680-70482680 on Build GRCh38
rs748655661 Chr.16: 70482735-70482735 on Build GRCh38
rs199686826 Chr.16: 70482762-70482762 on Build GRCh38
rs753577092 Chr.16: 70482678-70482678 on Build GRCh38
rs765074288 Chr.16: 70482774-70482774 on Build GRCh38
rs776249323 Chr.16: 70482846-70482846 on Build GRCh38
rs752657619 Chr.16: 70482782-70482782 on Build GRCh38

View Full Product Details