Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30955531-30955794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT14
Entrez Gene ID
79623
Gene Name
polypeptide N-acetylgalactosaminyltransferase 14
Gene Aliases
GALNT15, GalNac-T10, GalNac-T14
UniGene
Hs.468058
Gene Chromosome Location
Chr.2: 30888518-31138726 on Build GRCh38
Transcripts
NM_001253826.1 NM_001253827.1 NM_024572.3 XM_011533105.1 XM_017004906.1 XM_017004907.1 XM_017004908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7145153 c.607G>C p.E203Q Chr.2: 30955665-30955665 on Build GRCh38
3391698 c.592C>T p.L198F Chr.2: 30955680-30955680 on Build GRCh38
234732 c.636G>T p.L212L Chr.2: 30955636-30955636 on Build GRCh38
3391699 c.549G>A p.R183R Chr.2: 30955723-30955723 on Build GRCh38
1253043 c.588T>G p.T196T Chr.2: 30955684-30955684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999810829 Chr.2: 30955558-30955558 on Build GRCh38
rs766279646 Chr.2: 30955692-30955692 on Build GRCh38
rs35991873 Chr.2: 30955592-30955593 on Build GRCh38
rs150977129 Chr.2: 30955678-30955678 on Build GRCh38
rs368936056 Chr.2: 30955590-30955590 on Build GRCh38
rs41291165 Chr.2: 30955749-30955749 on Build GRCh38
rs149891806 Chr.2: 30955697-30955697 on Build GRCh38
rs200913085 Chr.2: 30955730-30955730 on Build GRCh38
rs772881413 Chr.2: 30955670-30955670 on Build GRCh38
rs528751269 Chr.2: 30955724-30955724 on Build GRCh38
rs747055588 Chr.2: 30955619-30955619 on Build GRCh38
rs765699135 Chr.2: 30955711-30955711 on Build GRCh38
rs747470499 Chr.2: 30955718-30955718 on Build GRCh38
rs76147880 Chr.2: 30955583-30955583 on Build GRCh38
rs767917819 Chr.2: 30955735-30955735 on Build GRCh38
rs762813718 Chr.2: 30955582-30955582 on Build GRCh38
rs367671327 Chr.2: 30955725-30955725 on Build GRCh38
rs200918843 Chr.2: 30955641-30955641 on Build GRCh38
rs763046385 Chr.2: 30955704-30955705 on Build GRCh38
rs774856872 Chr.2: 30955744-30955744 on Build GRCh38
rs776934740 Chr.2: 30955657-30955657 on Build GRCh38
rs145356432 Chr.2: 30955705-30955705 on Build GRCh38
rs774290160 Chr.2: 30955733-30955733 on Build GRCh38
rs757175498 Chr.2: 30955627-30955627 on Build GRCh38
rs780332326 Chr.2: 30955613-30955613 on Build GRCh38
rs772700119 Chr.2: 30955598-30955598 on Build GRCh38
rs73923311 Chr.2: 30955713-30955713 on Build GRCh38
rs780739128 Chr.2: 30955714-30955714 on Build GRCh38
rs142606595 Chr.2: 30955593-30955593 on Build GRCh38
rs773888092 Chr.2: 30955693-30955693 on Build GRCh38
rs148126284 Chr.2: 30955719-30955719 on Build GRCh38
rs1044159411 Chr.2: 30955710-30955710 on Build GRCh38
rs773106126 Chr.2: 30955729-30955729 on Build GRCh38
rs201069230 Chr.2: 30955567-30955567 on Build GRCh38
rs929547496 Chr.2: 30955662-30955662 on Build GRCh38
rs751054367 Chr.2: 30955585-30955585 on Build GRCh38
rs374183151 Chr.2: 30955586-30955586 on Build GRCh38
rs770668456 Chr.2: 30955731-30955731 on Build GRCh38
rs769639899 Chr.2: 30955666-30955666 on Build GRCh38
rs201974341 Chr.2: 30955760-30955760 on Build GRCh38
rs919529423 Chr.2: 30955660-30955660 on Build GRCh38
rs374214369 Chr.2: 30955676-30955677 on Build GRCh38
rs767057962 Chr.2: 30955742-30955742 on Build GRCh38
rs532750873 Chr.2: 30955629-30955629 on Build GRCh38
rs367877467 Chr.2: 30955571-30955571 on Build GRCh38
rs186978321 Chr.2: 30955561-30955561 on Build GRCh38
rs372237863 Chr.2: 30955592-30955592 on Build GRCh38
rs781070541 Chr.2: 30955721-30955721 on Build GRCh38
rs750157312 Chr.2: 30955709-30955709 on Build GRCh38
rs755655362 Chr.2: 30955706-30955706 on Build GRCh38

View Full Product Details