Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 140175896-140176123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLSTN2
Entrez Gene ID
64084
Gene Name
calsyntenin 2
Gene Aliases
ALC-GAMMA, CDHR13, CS2, CSTN2, alcagamma
UniGene
Hs.158529
Gene Chromosome Location
Chr.3: 139935185-140577397 on Build GRCh38
Transcripts
NM_022131.2 XM_017007022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1484679 c.145C>T p.H49Y Chr.3: 140175986-140175986 on Build GRCh38
3588105 c.180T>G p.I60M Chr.3: 140176021-140176021 on Build GRCh38
7239713 c.184G>T p.D62Y Chr.3: 140176025-140176025 on Build GRCh38
396022 c.154A>T p.I52L Chr.3: 140175995-140175995 on Build GRCh38
4979991 c.226T>G p.F76V Chr.3: 140176067-140176067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374979417 Chr.3: 140175949-140175949 on Build GRCh38
rs768540600 Chr.3: 140176062-140176062 on Build GRCh38
rs535846454 Chr.3: 140176010-140176010 on Build GRCh38
rs749946898 Chr.3: 140176059-140176059 on Build GRCh38
rs746878138 Chr.3: 140175960-140175960 on Build GRCh38
rs756644458 Chr.3: 140176031-140176031 on Build GRCh38
rs769802016 Chr.3: 140175996-140175996 on Build GRCh38
rs184749243 Chr.3: 140175973-140175973 on Build GRCh38
rs200330571 Chr.3: 140175958-140175958 on Build GRCh38
rs753058092 Chr.3: 140176027-140176027 on Build GRCh38
rs370618395 Chr.3: 140176085-140176085 on Build GRCh38
rs769869693 Chr.3: 140176086-140176086 on Build GRCh38
rs113636701 Chr.3: 140175988-140175988 on Build GRCh38
rs755991684 Chr.3: 140176001-140176001 on Build GRCh38
rs376445882 Chr.3: 140176060-140176060 on Build GRCh38
rs1045897392 Chr.3: 140175986-140175986 on Build GRCh38
rs751728404 Chr.3: 140176006-140176006 on Build GRCh38
rs768328479 Chr.3: 140175967-140175967 on Build GRCh38
rs1030107824 Chr.3: 140176093-140176093 on Build GRCh38
rs200226012 Chr.3: 140175994-140175994 on Build GRCh38
rs753756235 Chr.3: 140176076-140176076 on Build GRCh38
rs769577663 Chr.3: 140176087-140176087 on Build GRCh38
rs375577986 Chr.3: 140176092-140176092 on Build GRCh38
rs771716763 Chr.3: 140175957-140175957 on Build GRCh38
rs774684836 Chr.3: 140176094-140176094 on Build GRCh38
rs778186498 Chr.3: 140176040-140176040 on Build GRCh38
rs773271368 Chr.3: 140176089-140176089 on Build GRCh38
rs767751508 Chr.3: 140176014-140176014 on Build GRCh38
rs373766978 Chr.3: 140175998-140175998 on Build GRCh38
rs778417531 Chr.3: 140175936-140175936 on Build GRCh38
rs201489746 Chr.3: 140176090-140176090 on Build GRCh38
rs138350006 Chr.3: 140176102-140176102 on Build GRCh38
rs866552463 Chr.3: 140176046-140176046 on Build GRCh38

View Full Product Details