Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74588636-74588879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
-
UniGene
Hs.350775
Gene Chromosome Location
Chr.17: 74584679-74594209 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Gene Symbol
CD300LD
Entrez Gene ID
100131439
Gene Name
CD300 molecule like family member d
Gene Aliases
CD300D, CLM-4, CMRF35-A4, CMRF35A4
UniGene
Hs.531099
Gene Chromosome Location
Chr.17: 74579972-74592231 on Build GRCh38
Transcripts
NM_001302809.1 NM_152460.2 NM_001115152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521840 c.232C>T p.R78* Chr.17: 74588658-74588658 on Build GRCh38
4904931 c.147G>A p.L49L Chr.17: 74588743-74588743 on Build GRCh38
6352642 c.142T>A p.Y48N Chr.17: 74588748-74588748 on Build GRCh38
6556644 c.94C>A p.Q32K Chr.17: 74588796-74588796 on Build GRCh38
5444738 c.232C>A p.R78R Chr.17: 74588658-74588658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564832234 Chr.17: 74588736-74588736 on Build GRCh38
rs762229243 Chr.17: 74588676-74588676 on Build GRCh38
rs748383823 Chr.17: 74588820-74588820 on Build GRCh38
rs751143167 Chr.17: 74588694-74588694 on Build GRCh38
rs763733780 Chr.17: 74588793-74588793 on Build GRCh38
rs754428261 Chr.17: 74588703-74588703 on Build GRCh38
rs753724749 Chr.17: 74588797-74588797 on Build GRCh38
rs146929471 Chr.17: 74588857-74588857 on Build GRCh38
rs142719069 Chr.17: 74588748-74588748 on Build GRCh38
rs140321593 Chr.17: 74588728-74588730 on Build GRCh38
rs925736134 Chr.17: 74588824-74588824 on Build GRCh38
rs752361854 Chr.17: 74588791-74588791 on Build GRCh38
rs535714190 Chr.17: 74588711-74588711 on Build GRCh38
rs768796936 Chr.17: 74588858-74588858 on Build GRCh38
rs368705571 Chr.17: 74588749-74588749 on Build GRCh38
rs931441652 Chr.17: 74588842-74588842 on Build GRCh38
rs774693512 Chr.17: 74588776-74588776 on Build GRCh38
rs770203986 Chr.17: 74588835-74588835 on Build GRCh38
rs756980571 Chr.17: 74588800-74588800 on Build GRCh38
rs749707317 Chr.17: 74588840-74588840 on Build GRCh38
rs868497373 Chr.17: 74588750-74588750 on Build GRCh38
rs766967345 Chr.17: 74588787-74588787 on Build GRCh38
rs760255267 Chr.17: 74588792-74588792 on Build GRCh38
rs987422430 Chr.17: 74588781-74588781 on Build GRCh38
rs777015959 Chr.17: 74588669-74588669 on Build GRCh38
rs529615042 Chr.17: 74588836-74588836 on Build GRCh38
rs778949900 Chr.17: 74588801-74588801 on Build GRCh38
rs771521830 Chr.17: 74588762-74588762 on Build GRCh38
rs765806095 Chr.17: 74588691-74588691 on Build GRCh38
rs752310378 Chr.17: 74588716-74588716 on Build GRCh38
rs191085731 Chr.17: 74588724-74588724 on Build GRCh38
rs373955929 Chr.17: 74588686-74588686 on Build GRCh38
rs747426462 Chr.17: 74588812-74588814 on Build GRCh38
rs371226315 Chr.17: 74588721-74588721 on Build GRCh38
rs749188852 Chr.17: 74588733-74588733 on Build GRCh38
rs781360547 Chr.17: 74588812-74588812 on Build GRCh38
rs747280884 Chr.17: 74588809-74588809 on Build GRCh38
rs372269225 Chr.17: 74588782-74588782 on Build GRCh38
rs765739470 Chr.17: 74588677-74588677 on Build GRCh38
rs199533935 Chr.17: 74588738-74588738 on Build GRCh38
rs747063220 Chr.17: 74588658-74588658 on Build GRCh38
rs757286773 Chr.17: 74588734-74588734 on Build GRCh38
rs374599014 Chr.17: 74588747-74588747 on Build GRCh38
rs781234456 Chr.17: 74588735-74588735 on Build GRCh38
rs777023909 Chr.17: 74588739-74588739 on Build GRCh38
rs763576087 Chr.17: 74588689-74588689 on Build GRCh38
rs750286256 Chr.17: 74588804-74588804 on Build GRCh38

View Full Product Details