Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28392025-28392252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3CL
Entrez Gene ID
728689
Gene Name
eukaryotic translation initiation factor 3 subunit C-like
Gene Aliases
-
UniGene
Hs.567374
Gene Chromosome Location
Chr.16: 28379581-28403885 on Build GRCh38
Transcripts
NM_001099661.2 NM_001317856.1 NM_001317857.1 XM_017023620.1 XM_017023621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430215 c.859A>G p.K287E Chr.16: 28392060-28392060 on Build GRCh38
6880823 c.808G>A p.A270T Chr.16: 28392111-28392111 on Build GRCh38
6374657 c.876G>T p.E292D Chr.16: 28392043-28392043 on Build GRCh38
6191146 c.871_873delGAG p.E295delE Chr.16: 28392046-28392048 on Build GRCh38
1251082 c.882_887delGGAGGA p.E294_E295delEE Chr.16: 28392032-28392037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769927523 Chr.16: 28392111-28392111 on Build GRCh38
rs750287215 Chr.16: 28392086-28392086 on Build GRCh38
rs369338836 Chr.16: 28392144-28392144 on Build GRCh38
rs553397106 Chr.16: 28392110-28392110 on Build GRCh38
rs555978476 Chr.16: 28392150-28392150 on Build GRCh38
rs781624187 Chr.16: 28392100-28392100 on Build GRCh38
rs948741899 Chr.16: 28392166-28392166 on Build GRCh38
rs915873435 Chr.16: 28392163-28392163 on Build GRCh38
rs535752459 Chr.16: 28392146-28392146 on Build GRCh38
rs758248692 Chr.16: 28392099-28392099 on Build GRCh38
rs748248736 Chr.16: 28392109-28392109 on Build GRCh38
rs374727655 Chr.16: 28392073-28392073 on Build GRCh38
rs981522220 Chr.16: 28392172-28392172 on Build GRCh38

View Full Product Details