Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 84619427-84619660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS1
Entrez Gene ID
1040
Gene Name
CDP-diacylglycerol synthase 1
Gene Aliases
CDS
UniGene
Hs.654899
Gene Chromosome Location
Chr.4: 84582904-84651340 on Build GRCh38
Transcripts
NM_001263.3 XM_005262687.2 XM_017007648.1 XM_017007649.1 XM_017007650.1 XM_017007651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625803 c.542G>A p.R181H Chr.4: 84619495-84619495 on Build GRCh38
4499101 c.534C>T p.F178F Chr.4: 84619487-84619487 on Build GRCh38
1058329 c.540T>A p.I180I Chr.4: 84619493-84619493 on Build GRCh38
7429111 c.505T>C p.F169L Chr.4: 84619458-84619458 on Build GRCh38
5807599 c.525A>T p.Q175H Chr.4: 84619478-84619478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772501123 Chr.4: 84619567-84619567 on Build GRCh38
rs34639898 Chr.4: 84619462-84619463 on Build GRCh38
rs111333888 Chr.4: 84619588-84619588 on Build GRCh38
rs1028154893 Chr.4: 84619621-84619621 on Build GRCh38
rs759347542 Chr.4: 84619494-84619494 on Build GRCh38
rs992852332 Chr.4: 84619611-84619611 on Build GRCh38
rs779491131 Chr.4: 84619559-84619559 on Build GRCh38
rs959634224 Chr.4: 84619600-84619600 on Build GRCh38
rs961849659 Chr.4: 84619623-84619623 on Build GRCh38
rs748535874 Chr.4: 84619562-84619562 on Build GRCh38
rs190844825 Chr.4: 84619548-84619548 on Build GRCh38
rs1025141197 Chr.4: 84619613-84619613 on Build GRCh38
rs143213768 Chr.4: 84619518-84619518 on Build GRCh38
rs773220033 Chr.4: 84619454-84619454 on Build GRCh38
rs764952660 Chr.4: 84619495-84619495 on Build GRCh38
rs201393477 Chr.4: 84619555-84619555 on Build GRCh38
rs760782069 Chr.4: 84619478-84619478 on Build GRCh38
rs754016381 Chr.4: 84619490-84619490 on Build GRCh38
rs755400754 Chr.4: 84619558-84619558 on Build GRCh38
rs781015119 Chr.4: 84619553-84619553 on Build GRCh38
rs751071726 Chr.4: 84619526-84619526 on Build GRCh38
rs766546783 Chr.4: 84619484-84619484 on Build GRCh38
rs368014358 Chr.4: 84619572-84619572 on Build GRCh38
rs1016656370 Chr.4: 84619525-84619525 on Build GRCh38
rs144762911 Chr.4: 84619521-84619521 on Build GRCh38
rs747087647 Chr.4: 84619579-84619579 on Build GRCh38
rs752635583 Chr.4: 84619505-84619505 on Build GRCh38

View Full Product Details