Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154402586-154402811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L1
Entrez Gene ID
1774
Gene Name
deoxyribonuclease I-like 1
Gene Aliases
DNAS1L1, DNASEX, DNL1L, G4.8, XIB
UniGene
Hs.401929
Gene Chromosome Location
Chr.X: 154401236-154412101 on Build GRCh38
Transcripts
NM_001009932.2 NM_001009933.2 NM_001009934.2 NM_001303620.1 NM_006730.3 XM_005277829.4 XM_011531122.2 XM_017029332.1

SNP Detail

SNP ID Chromosome Location
rs369393190 Chr.X: 154402718-154402718 on Build GRCh38
rs781969793 Chr.X: 154402685-154402685 on Build GRCh38
rs782422883 Chr.X: 154402611-154402611 on Build GRCh38
rs201589296 Chr.X: 154402689-154402689 on Build GRCh38
rs781988297 Chr.X: 154402641-154402641 on Build GRCh38
rs72616477 Chr.X: 154402692-154402692 on Build GRCh38
rs781924417 Chr.X: 154402616-154402616 on Build GRCh38
rs887920880 Chr.X: 154402703-154402703 on Build GRCh38
rs782047097 Chr.X: 154402733-154402733 on Build GRCh38
rs781964455 Chr.X: 154402727-154402727 on Build GRCh38
rs146004451 Chr.X: 154402789-154402789 on Build GRCh38
rs782790575 Chr.X: 154402760-154402760 on Build GRCh38
rs782050602 Chr.X: 154402688-154402689 on Build GRCh38
rs782282210 Chr.X: 154402606-154402606 on Build GRCh38
rs781833694 Chr.X: 154402786-154402786 on Build GRCh38
rs782416192 Chr.X: 154402698-154402698 on Build GRCh38
rs782526750 Chr.X: 154402679-154402679 on Build GRCh38
rs377616504 Chr.X: 154402661-154402661 on Build GRCh38
rs781860370 Chr.X: 154402676-154402676 on Build GRCh38
rs782275215 Chr.X: 154402695-154402695 on Build GRCh38
rs782052899 Chr.X: 154402700-154402701 on Build GRCh38
rs782351883 Chr.X: 154402720-154402720 on Build GRCh38
rs782263838 Chr.X: 154402705-154402705 on Build GRCh38
rs138207690 Chr.X: 154402704-154402704 on Build GRCh38
rs782259110 Chr.X: 154402647-154402647 on Build GRCh38
rs187477756 Chr.X: 154402648-154402648 on Build GRCh38
rs782436955 Chr.X: 154402690-154402690 on Build GRCh38
rs17849495 Chr.X: 154402785-154402785 on Build GRCh38

View Full Product Details