Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17613154-17613389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1E1
Entrez Gene ID
529
Gene Name
ATPase H+ transporting V1 subunit E1
Gene Aliases
ATP6E, ATP6E2, ATP6V1E, P31, Vma4
UniGene
Hs.517338
Gene Chromosome Location
Chr.22: 17592136-17628822 on Build GRCh38
Transcripts
NM_001039366.1 NM_001039367.1 NM_001696.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
33399 c.148A>G p.R50G Chr.22: 17613272-17613272 on Build GRCh38
4520680 c.106G>A p.E36K Chr.22: 17613314-17613314 on Build GRCh38
1031874 c.195G>A p.E65E Chr.22: 17613225-17613225 on Build GRCh38
4642586 c.134T>C p.L45P Chr.22: 17613286-17613286 on Build GRCh38
6236931 c.131G>A p.R44Q Chr.22: 17613289-17613289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749946288 Chr.22: 17613266-17613266 on Build GRCh38
rs755637338 Chr.22: 17613274-17613274 on Build GRCh38
rs748514222 Chr.22: 17613335-17613335 on Build GRCh38
rs199646306 Chr.22: 17613191-17613191 on Build GRCh38
rs753477779 Chr.22: 17613290-17613290 on Build GRCh38
rs976952211 Chr.22: 17613347-17613347 on Build GRCh38
rs755341097 Chr.22: 17613181-17613181 on Build GRCh38
rs774153075 Chr.22: 17613249-17613249 on Build GRCh38
rs369896095 Chr.22: 17613183-17613183 on Build GRCh38
rs779373750 Chr.22: 17613303-17613303 on Build GRCh38
rs376703987 Chr.22: 17613204-17613204 on Build GRCh38
rs772664694 Chr.22: 17613357-17613358 on Build GRCh38
rs964434989 Chr.22: 17613239-17613239 on Build GRCh38
rs1021709800 Chr.22: 17613360-17613360 on Build GRCh38
rs370760245 Chr.22: 17613197-17613197 on Build GRCh38
rs761786548 Chr.22: 17613261-17613261 on Build GRCh38
rs373352619 Chr.22: 17613188-17613188 on Build GRCh38
rs767395917 Chr.22: 17613262-17613262 on Build GRCh38
rs754712901 Chr.22: 17613299-17613299 on Build GRCh38
rs138309443 Chr.22: 17613228-17613228 on Build GRCh38
rs771593342 Chr.22: 17613190-17613191 on Build GRCh38
rs766001041 Chr.22: 17613289-17613289 on Build GRCh38
rs956118032 Chr.22: 17613277-17613277 on Build GRCh38
rs11544402 Chr.22: 17613275-17613275 on Build GRCh38
rs376066633 Chr.22: 17613346-17613346 on Build GRCh38
rs758937100 Chr.22: 17613339-17613339 on Build GRCh38
rs763836312 Chr.22: 17613229-17613229 on Build GRCh38
rs1029828132 Chr.22: 17613348-17613348 on Build GRCh38
rs776773108 Chr.22: 17613219-17613219 on Build GRCh38

View Full Product Details