Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246641832-246642079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNST
Entrez Gene ID
163882
Gene Name
consortin, connexin sorting protein
Gene Aliases
C1orf71, PPP1R64
UniGene
Hs.368353
Gene Chromosome Location
Chr.1: 246566337-246668587 on Build GRCh38
Transcripts
NM_001139459.1 NM_152609.2 XM_005273081.3 XM_005273083.4 XM_011544110.2 XM_011544111.1 XM_011544112.1 XM_011544113.1 XM_011544114.2 XM_017000487.1 XM_017000488.1 XM_017000489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846081 c.892G>C p.V298L Chr.1: 246641992-246641992 on Build GRCh38
1473740 c.909G>C p.K303N Chr.1: 246642009-246642009 on Build GRCh38
7352386 c.863A>G p.Q288R Chr.1: 246641963-246641963 on Build GRCh38
1190707 c.870delG p.K291fs*7 Chr.1: 246641970-246641970 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755112081 Chr.1: 246641920-246641922 on Build GRCh38
rs1031346607 Chr.1: 246642024-246642024 on Build GRCh38
rs902060173 Chr.1: 246641873-246641873 on Build GRCh38
rs997641940 Chr.1: 246641882-246641882 on Build GRCh38
rs992076403 Chr.1: 246642050-246642050 on Build GRCh38
rs1021644109 Chr.1: 246641980-246641980 on Build GRCh38
rs993186411 Chr.1: 246641886-246641886 on Build GRCh38
rs756826028 Chr.1: 246642052-246642052 on Build GRCh38
rs895770891 Chr.1: 246641928-246641928 on Build GRCh38
rs138732495 Chr.1: 246641950-246641950 on Build GRCh38
rs141796133 Chr.1: 246641968-246641968 on Build GRCh38
rs1026360505 Chr.1: 246641908-246641908 on Build GRCh38
rs746206552 Chr.1: 246641961-246641961 on Build GRCh38
rs756880893 Chr.1: 246641920-246641920 on Build GRCh38
rs776833324 Chr.1: 246641985-246641985 on Build GRCh38
rs753366028 Chr.1: 246641913-246641913 on Build GRCh38
rs760555659 Chr.1: 246642028-246642028 on Build GRCh38
rs756047402 Chr.1: 246641909-246641909 on Build GRCh38
rs373967697 Chr.1: 246641982-246641982 on Build GRCh38
rs148471784 Chr.1: 246642004-246642004 on Build GRCh38
rs369755986 Chr.1: 246641981-246641981 on Build GRCh38
rs752462986 Chr.1: 246642017-246642017 on Build GRCh38
rs764065365 Chr.1: 246642030-246642030 on Build GRCh38
rs781497701 Chr.1: 246641989-246641989 on Build GRCh38
rs375376106 Chr.1: 246641911-246641911 on Build GRCh38
rs763094160 Chr.1: 246642002-246642002 on Build GRCh38
rs753314802 Chr.1: 246642042-246642042 on Build GRCh38
rs535392402 Chr.1: 246641997-246641997 on Build GRCh38
rs770140290 Chr.1: 246641991-246641991 on Build GRCh38
rs761842047 Chr.1: 246642040-246642040 on Build GRCh38
rs762028751 Chr.1: 246641989-246641989 on Build GRCh38
rs773537941 Chr.1: 246641993-246641993 on Build GRCh38
rs184541472 Chr.1: 246641895-246641895 on Build GRCh38
rs115633389 Chr.1: 246641963-246641963 on Build GRCh38
rs372047884 Chr.1: 246641894-246641894 on Build GRCh38
rs960619286 Chr.1: 246642043-246642043 on Build GRCh38

View Full Product Details