Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 33875824-33876085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVEN
Entrez Gene ID
57099
Gene Name
apoptosis and caspase activation inhibitor
Gene Aliases
PDCD12
UniGene
Hs.555966
Gene Chromosome Location
Chr.15: 33858602-34074877 on Build GRCh38
Transcripts
NM_020371.2 XM_011521818.2 XM_011521819.2 XM_011521820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351288 c.516+10G>C p.? Chr.15: 33875915-33875915 on Build GRCh38
7341138 c.473C>T p.A158V Chr.15: 33875968-33875968 on Build GRCh38
6326818 c.475G>A p.E159K Chr.15: 33875966-33875966 on Build GRCh38
3500505 c.467G>A p.R156Q Chr.15: 33875974-33875974 on Build GRCh38
7248551 c.447G>T p.G149G Chr.15: 33875994-33875994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982571572 Chr.15: 33876017-33876017 on Build GRCh38
rs200947045 Chr.15: 33876008-33876008 on Build GRCh38
rs754556886 Chr.15: 33875885-33875885 on Build GRCh38
rs755279877 Chr.15: 33875906-33875908 on Build GRCh38
rs779420752 Chr.15: 33875876-33875876 on Build GRCh38
rs999026023 Chr.15: 33876019-33876019 on Build GRCh38
rs6495303 Chr.15: 33876036-33876036 on Build GRCh38
rs772158926 Chr.15: 33875933-33875933 on Build GRCh38
rs11295548 Chr.15: 33876007-33876007 on Build GRCh38
rs764388449 Chr.15: 33875980-33875980 on Build GRCh38
rs765955171 Chr.15: 33875995-33875995 on Build GRCh38
rs760432883 Chr.15: 33876007-33876009 on Build GRCh38
rs201845444 Chr.15: 33876007-33876007 on Build GRCh38
rs144530675 Chr.15: 33875888-33875888 on Build GRCh38
rs973064672 Chr.15: 33875934-33875934 on Build GRCh38
rs372196341 Chr.15: 33875883-33875883 on Build GRCh38
rs10519888 Chr.15: 33875915-33875915 on Build GRCh38
rs146625663 Chr.15: 33875929-33875929 on Build GRCh38
rs1023324277 Chr.15: 33876049-33876049 on Build GRCh38
rs889095921 Chr.15: 33875938-33875938 on Build GRCh38
rs772097092 Chr.15: 33876040-33876040 on Build GRCh38
rs1027598001 Chr.15: 33875975-33875975 on Build GRCh38
rs747981234 Chr.15: 33875890-33875890 on Build GRCh38
rs557308961 Chr.15: 33876020-33876020 on Build GRCh38
rs770738383 Chr.15: 33876024-33876024 on Build GRCh38
rs773888669 Chr.15: 33875936-33875936 on Build GRCh38
rs969448249 Chr.15: 33876051-33876051 on Build GRCh38
rs945005063 Chr.15: 33876003-33876003 on Build GRCh38
rs546781863 Chr.15: 33875926-33875926 on Build GRCh38
rs763787086 Chr.15: 33876020-33876020 on Build GRCh38
rs549272762 Chr.15: 33875863-33875863 on Build GRCh38
rs771856187 Chr.15: 33876007-33876008 on Build GRCh38
rs769670280 Chr.15: 33875913-33875913 on Build GRCh38
rs1003134906 Chr.15: 33876061-33876061 on Build GRCh38
rs1050631942 Chr.15: 33876058-33876058 on Build GRCh38
rs769979964 Chr.15: 33876004-33876005 on Build GRCh38
rs376772386 Chr.15: 33875884-33875886 on Build GRCh38
rs530806670 Chr.15: 33875859-33875859 on Build GRCh38
rs144646195 Chr.15: 33875989-33875989 on Build GRCh38
rs1029562144 Chr.15: 33875903-33875903 on Build GRCh38
rs1009037838 Chr.15: 33875909-33875909 on Build GRCh38
rs867197650 Chr.15: 33875898-33875898 on Build GRCh38
rs751012201 Chr.15: 33876005-33876005 on Build GRCh38
rs749177881 Chr.15: 33875919-33875919 on Build GRCh38
rs890224152 Chr.15: 33875906-33875906 on Build GRCh38
rs372785565 Chr.15: 33875974-33875974 on Build GRCh38
rs749314882 Chr.15: 33876021-33876021 on Build GRCh38
rs748285917 Chr.15: 33875966-33875972 on Build GRCh38
rs996323814 Chr.15: 33875894-33875894 on Build GRCh38
rs958601135 Chr.15: 33875983-33875983 on Build GRCh38
rs755985571 Chr.15: 33876009-33876009 on Build GRCh38
rs796673242 Chr.15: 33876019-33876019 on Build GRCh38
rs745821622 Chr.15: 33876006-33876007 on Build GRCh38
rs761035754 Chr.15: 33875979-33875979 on Build GRCh38
rs754404649 Chr.15: 33875984-33875984 on Build GRCh38
rs781531896 Chr.15: 33875906-33875910 on Build GRCh38

View Full Product Details