Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65919519-65919762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAP1
Entrez Gene ID
10589
Gene Name
DR1 associated protein 1
Gene Aliases
NC2-alpha
UniGene
Hs.356742
Gene Chromosome Location
Chr.11: 65919257-65921577 on Build GRCh38
Transcripts
NM_006442.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6571252 c.36C>T p.F12F Chr.11: 65919537-65919537 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs954943734 Chr.11: 65919619-65919619 on Build GRCh38
rs778203628 Chr.11: 65919594-65919595 on Build GRCh38
rs939978751 Chr.11: 65919552-65919552 on Build GRCh38
rs981730753 Chr.11: 65919634-65919634 on Build GRCh38
rs550152736 Chr.11: 65919629-65919629 on Build GRCh38
rs907766101 Chr.11: 65919679-65919679 on Build GRCh38
rs761156509 Chr.11: 65919551-65919551 on Build GRCh38
rs989548505 Chr.11: 65919670-65919670 on Build GRCh38
rs938645985 Chr.11: 65919702-65919702 on Build GRCh38
rs530128982 Chr.11: 65919569-65919569 on Build GRCh38
rs111481652 Chr.11: 65919545-65919545 on Build GRCh38
rs752879700 Chr.11: 65919729-65919729 on Build GRCh38
rs964680635 Chr.11: 65919665-65919665 on Build GRCh38
rs959114306 Chr.11: 65919655-65919656 on Build GRCh38
rs1028905476 Chr.11: 65919574-65919574 on Build GRCh38
rs894169164 Chr.11: 65919718-65919718 on Build GRCh38
rs764080948 Chr.11: 65919735-65919735 on Build GRCh38
rs751719901 Chr.11: 65919738-65919738 on Build GRCh38
rs1041056167 Chr.11: 65919684-65919684 on Build GRCh38
rs893942672 Chr.11: 65919590-65919590 on Build GRCh38
rs552385153 Chr.11: 65919692-65919692 on Build GRCh38
rs563853124 Chr.11: 65919672-65919672 on Build GRCh38
rs1034682123 Chr.11: 65919641-65919641 on Build GRCh38
rs1021571377 Chr.11: 65919728-65919728 on Build GRCh38
rs1017592382 Chr.11: 65919656-65919656 on Build GRCh38
rs987439307 Chr.11: 65919626-65919626 on Build GRCh38
rs772845646 Chr.11: 65919543-65919543 on Build GRCh38
rs920346240 Chr.11: 65919667-65919667 on Build GRCh38
rs1011059028 Chr.11: 65919718-65919719 on Build GRCh38
rs967389905 Chr.11: 65919632-65919633 on Build GRCh38
rs990491598 Chr.11: 65919663-65919663 on Build GRCh38
rs112677209 Chr.11: 65919540-65919540 on Build GRCh38
rs867958994 Chr.11: 65919657-65919657 on Build GRCh38
rs758440372 Chr.11: 65919730-65919730 on Build GRCh38
rs924529986 Chr.11: 65919694-65919694 on Build GRCh38
rs534480581 Chr.11: 65919740-65919740 on Build GRCh38
rs1026999025 Chr.11: 65919598-65919598 on Build GRCh38
rs565714575 Chr.11: 65919704-65919704 on Build GRCh38
rs765391343 Chr.11: 65919571-65919571 on Build GRCh38
rs944633306 Chr.11: 65919682-65919682 on Build GRCh38
rs369509341 Chr.11: 65919582-65919582 on Build GRCh38
rs532491202 Chr.11: 65919686-65919686 on Build GRCh38

View Full Product Details