Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14908764-14909035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1C
Entrez Gene ID
64421
Gene Name
DNA cross-link repair 1C
Gene Aliases
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
UniGene
Hs.655932
Gene Chromosome Location
Chr.10: 14897853-14954432 on Build GRCh38
Transcripts
NM_001033855.2 NM_001033857.2 NM_001033858.2 NM_001289076.1 NM_001289077.1 NM_001289078.1 NM_001289079.1 NM_022487.3 NR_110297.1 XM_006717491.3 XM_011519616.1 XM_011519617.1 XM_011519619.1 XM_017016556.1 XM_017016557.1 XM_017016558.1 XM_017016559.1 XM_017016560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3436134 c.1321G>A p.D441N Chr.10: 14908821-14908821 on Build GRCh38
4651769 c.1174T>G p.F392V Chr.10: 14908968-14908968 on Build GRCh38
4385964 c.1700_1701insAACA p.S567fs*12 Chr.10: 14908786-14908787 on Build GRCh38
916629 c.1126G>T p.V376L Chr.10: 14909016-14909016 on Build GRCh38
4333117 c.1458A>G p.V486V Chr.10: 14909029-14909029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750613031 Chr.10: 14909008-14909009 on Build GRCh38
rs766549429 Chr.10: 14908831-14908831 on Build GRCh38
rs766778472 Chr.10: 14908924-14908924 on Build GRCh38
rs1023508705 Chr.10: 14908942-14908943 on Build GRCh38
rs772512363 Chr.10: 14908910-14908910 on Build GRCh38
rs41300670 Chr.10: 14908984-14908984 on Build GRCh38
rs867997110 Chr.10: 14908947-14908947 on Build GRCh38
rs1029319156 Chr.10: 14908882-14908882 on Build GRCh38
rs780875686 Chr.10: 14908983-14908983 on Build GRCh38
rs750692726 Chr.10: 14908817-14908817 on Build GRCh38
rs1017166380 Chr.10: 14908897-14908897 on Build GRCh38
rs138799247 Chr.10: 14908923-14908923 on Build GRCh38
rs886043100 Chr.10: 14908825-14908825 on Build GRCh38
rs752378965 Chr.10: 14908847-14908847 on Build GRCh38
rs762293150 Chr.10: 14908788-14908788 on Build GRCh38
rs777250271 Chr.10: 14908860-14908860 on Build GRCh38
rs1013045171 Chr.10: 14908934-14908934 on Build GRCh38
rs777600117 Chr.10: 14908960-14908960 on Build GRCh38
rs780648720 Chr.10: 14908874-14908874 on Build GRCh38
rs756239667 Chr.10: 14909009-14909011 on Build GRCh38
rs887381468 Chr.10: 14908883-14908883 on Build GRCh38
rs767573884 Chr.10: 14908795-14908795 on Build GRCh38
rs41300672 Chr.10: 14908942-14908942 on Build GRCh38
rs114767681 Chr.10: 14908996-14908996 on Build GRCh38
rs760791470 Chr.10: 14908830-14908830 on Build GRCh38
rs773500841 Chr.10: 14908921-14908921 on Build GRCh38
rs963209409 Chr.10: 14908928-14908928 on Build GRCh38
rs921962593 Chr.10: 14908899-14908899 on Build GRCh38
rs751349938 Chr.10: 14908963-14908963 on Build GRCh38
rs756895191 Chr.10: 14908981-14908981 on Build GRCh38
rs542791233 Chr.10: 14908931-14908931 on Build GRCh38
rs41300674 Chr.10: 14908864-14908864 on Build GRCh38
rs752712893 Chr.10: 14908946-14908946 on Build GRCh38
rs779242343 Chr.10: 14908993-14908993 on Build GRCh38
rs755622031 Chr.10: 14908986-14908986 on Build GRCh38
rs754963036 Chr.10: 14908833-14908833 on Build GRCh38
rs767758218 Chr.10: 14908855-14908859 on Build GRCh38
rs779050968 Chr.10: 14908836-14908836 on Build GRCh38
rs769134680 Chr.10: 14908892-14908892 on Build GRCh38
rs61745540 Chr.10: 14908885-14908885 on Build GRCh38
rs372597855 Chr.10: 14908995-14908995 on Build GRCh38
rs529177486 Chr.10: 14908900-14908900 on Build GRCh38
rs762013968 Chr.10: 14908906-14908906 on Build GRCh38
rs183622528 Chr.10: 14908851-14908851 on Build GRCh38
rs758077996 Chr.10: 14908952-14908952 on Build GRCh38
rs753955796 Chr.10: 14908927-14908927 on Build GRCh38
rs886037924 Chr.10: 14908817-14908818 on Build GRCh38
rs754013987 Chr.10: 14908832-14908832 on Build GRCh38

View Full Product Details