Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100678468-100678693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB2
Entrez Gene ID
2783
Gene Name
G protein subunit beta 2
Gene Aliases
-
UniGene
Hs.185172
Gene Chromosome Location
Chr.7: 100673740-100679169 on Build GRCh38
Transcripts
NM_005273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3928671 c.910C>A p.R304S Chr.7: 100678608-100678608 on Build GRCh38
5459281 c.916+9T>C p.? Chr.7: 100678623-100678623 on Build GRCh38
4153677 c.791A>T p.Y264F Chr.7: 100678489-100678489 on Build GRCh38
6253861 c.859G>T p.A287S Chr.7: 100678557-100678557 on Build GRCh38
4649028 c.853C>T p.L285L Chr.7: 100678551-100678551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765996409 Chr.7: 100678536-100678536 on Build GRCh38
rs200568621 Chr.7: 100678658-100678658 on Build GRCh38
rs754445772 Chr.7: 100678538-100678538 on Build GRCh38
rs775439910 Chr.7: 100678494-100678494 on Build GRCh38
rs747216923 Chr.7: 100678657-100678657 on Build GRCh38
rs768831809 Chr.7: 100678574-100678574 on Build GRCh38
rs1046454557 Chr.7: 100678626-100678626 on Build GRCh38
rs938714830 Chr.7: 100678594-100678594 on Build GRCh38
rs199877757 Chr.7: 100678662-100678662 on Build GRCh38
rs757042963 Chr.7: 100678655-100678655 on Build GRCh38
rs147810006 Chr.7: 100678501-100678501 on Build GRCh38
rs755961772 Chr.7: 100678550-100678550 on Build GRCh38
rs142525639 Chr.7: 100678565-100678565 on Build GRCh38
rs772265632 Chr.7: 100678490-100678490 on Build GRCh38
rs757495640 Chr.7: 100678568-100678568 on Build GRCh38
rs151109351 Chr.7: 100678577-100678577 on Build GRCh38
rs202102462 Chr.7: 100678505-100678505 on Build GRCh38
rs777710481 Chr.7: 100678647-100678647 on Build GRCh38
rs752609096 Chr.7: 100678547-100678547 on Build GRCh38
rs756013914 Chr.7: 100678634-100678634 on Build GRCh38
rs750156691 Chr.7: 100678529-100678529 on Build GRCh38
rs773728037 Chr.7: 100678665-100678665 on Build GRCh38
rs574729024 Chr.7: 100678620-100678620 on Build GRCh38
rs762404805 Chr.7: 100678593-100678593 on Build GRCh38
rs781425748 Chr.7: 100678660-100678660 on Build GRCh38
rs148398821 Chr.7: 100678541-100678541 on Build GRCh38
rs781337161 Chr.7: 100678622-100678622 on Build GRCh38
rs1057139137 Chr.7: 100678617-100678617 on Build GRCh38
rs201331306 Chr.7: 100678537-100678537 on Build GRCh38
rs777430598 Chr.7: 100678551-100678551 on Build GRCh38
rs367625456 Chr.7: 100678666-100678666 on Build GRCh38
rs757991795 Chr.7: 100678535-100678535 on Build GRCh38
rs996279616 Chr.7: 100678664-100678664 on Build GRCh38
rs199609383 Chr.7: 100678624-100678624 on Build GRCh38
rs370708626 Chr.7: 100678559-100678559 on Build GRCh38
rs530703465 Chr.7: 100678610-100678610 on Build GRCh38
rs370617239 Chr.7: 100678630-100678630 on Build GRCh38
rs753600090 Chr.7: 100678649-100678649 on Build GRCh38
rs772984234 Chr.7: 100678498-100678500 on Build GRCh38
rs762313075 Chr.7: 100678514-100678514 on Build GRCh38
rs570401973 Chr.7: 100678500-100678500 on Build GRCh38
rs200286640 Chr.7: 100678604-100678604 on Build GRCh38
rs760500319 Chr.7: 100678628-100678628 on Build GRCh38
rs145480659 Chr.7: 100678556-100678556 on Build GRCh38
rs376361254 Chr.7: 100678605-100678605 on Build GRCh38
rs893589422 Chr.7: 100678668-100678668 on Build GRCh38
rs770268754 Chr.7: 100678596-100678596 on Build GRCh38
rs977785007 Chr.7: 100678495-100678495 on Build GRCh38
rs771306949 Chr.7: 100678667-100678667 on Build GRCh38
rs748181605 Chr.7: 100678661-100678661 on Build GRCh38
rs765970588 Chr.7: 100678655-100678656 on Build GRCh38
rs765502920 Chr.7: 100678521-100678521 on Build GRCh38

View Full Product Details