Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 94478772-94479041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCT
Entrez Gene ID
1638
Gene Name
dopachrome tautomerase
Gene Aliases
TRP-2, TYRP2
UniGene
Hs.301865
Gene Chromosome Location
Chr.13: 94437304-94550265 on Build GRCh38
Transcripts
NM_001129889.2 NM_001322182.1 NM_001322183.1 NM_001322184.1 NM_001322185.1 NM_001322186.1 NM_001922.4 XM_011521049.1 XM_017020401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721428 c.248G>A p.R83H Chr.13: 94479008-94479008 on Build GRCh38
98357 c.248G>A p.R83H Chr.13: 94479008-94479008 on Build GRCh38
6227382 c.269T>G p.F90C Chr.13: 94478987-94478987 on Build GRCh38
5682404 c.238C>G p.Q80E Chr.13: 94479018-94479018 on Build GRCh38
7266870 c.267A>T p.K89N Chr.13: 94478989-94478989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771623487 Chr.13: 94478910-94478910 on Build GRCh38
rs770498870 Chr.13: 94478926-94478926 on Build GRCh38
rs61758413 Chr.13: 94478828-94478828 on Build GRCh38
rs369559246 Chr.13: 94478942-94478942 on Build GRCh38
rs1029103096 Chr.13: 94478804-94478804 on Build GRCh38
rs750723500 Chr.13: 94478948-94478948 on Build GRCh38
rs961100239 Chr.13: 94478890-94478890 on Build GRCh38
rs771963278 Chr.13: 94478989-94478989 on Build GRCh38
rs543465906 Chr.13: 94478872-94478872 on Build GRCh38
rs550904509 Chr.13: 94478929-94478929 on Build GRCh38
rs951706722 Chr.13: 94478810-94478810 on Build GRCh38
rs760169154 Chr.13: 94478921-94478921 on Build GRCh38
rs756231813 Chr.13: 94478951-94478951 on Build GRCh38
rs767647386 Chr.13: 94478945-94478945 on Build GRCh38
rs374657624 Chr.13: 94479008-94479008 on Build GRCh38
rs975223665 Chr.13: 94478898-94478898 on Build GRCh38
rs762029610 Chr.13: 94478944-94478944 on Build GRCh38
rs762079242 Chr.13: 94479015-94479015 on Build GRCh38
rs982668325 Chr.13: 94478848-94478848 on Build GRCh38
rs529034525 Chr.13: 94478937-94478937 on Build GRCh38
rs778698665 Chr.13: 94478979-94478979 on Build GRCh38
rs1014794831 Chr.13: 94478824-94478824 on Build GRCh38
rs747904928 Chr.13: 94478985-94478985 on Build GRCh38
rs1006133750 Chr.13: 94478815-94478815 on Build GRCh38
rs780328275 Chr.13: 94478962-94478962 on Build GRCh38
rs772716942 Chr.13: 94478918-94478918 on Build GRCh38
rs61758412 Chr.13: 94478836-94478836 on Build GRCh38
rs777291106 Chr.13: 94478996-94478996 on Build GRCh38
rs774719717 Chr.13: 94479010-94479010 on Build GRCh38
rs764471543 Chr.13: 94478941-94478941 on Build GRCh38
rs199664123 Chr.13: 94479007-94479007 on Build GRCh38
rs763411727 Chr.13: 94479009-94479009 on Build GRCh38
rs928462472 Chr.13: 94478882-94478882 on Build GRCh38
rs995920707 Chr.13: 94478800-94478800 on Build GRCh38
rs577582856 Chr.13: 94478866-94478866 on Build GRCh38
rs151208955 Chr.13: 94478978-94478978 on Build GRCh38
rs912409319 Chr.13: 94478851-94478851 on Build GRCh38
rs981694409 Chr.13: 94478829-94478829 on Build GRCh38
rs753777021 Chr.13: 94478968-94478968 on Build GRCh38

View Full Product Details