Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46265018-46265267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR3
Entrez Gene ID
1232
Gene Name
C-C motif chemokine receptor 3
Gene Aliases
CC-CKR-3, CD193, CKR3, CMKBR3
UniGene
Hs.506190
Gene Chromosome Location
Chr.3: 46210699-46266706 on Build GRCh38
Transcripts
NM_001164680.1 NM_001837.3 NM_178328.1 NM_178329.2 XM_006712960.3 XM_011533335.2 XM_017005685.1 XM_017005686.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623204 c.8C>T p.T3I Chr.3: 46265166-46265166 on Build GRCh38
4832615 c.74C>T p.S25L Chr.3: 46265169-46265169 on Build GRCh38
347263 c.57C>T p.D19D Chr.3: 46265215-46265215 on Build GRCh38
3068011 c.58G>A p.V20M Chr.3: 46265216-46265216 on Build GRCh38
6623203 c.71C>T p.T24I Chr.3: 46265166-46265166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935219079 Chr.3: 46265176-46265176 on Build GRCh38
rs947476653 Chr.3: 46265106-46265106 on Build GRCh38
rs972634551 Chr.3: 46265065-46265065 on Build GRCh38
rs780698715 Chr.3: 46265187-46265187 on Build GRCh38
rs767403282 Chr.3: 46265211-46265211 on Build GRCh38
rs535223919 Chr.3: 46265096-46265096 on Build GRCh38
rs200354348 Chr.3: 46265166-46265166 on Build GRCh38
rs755271586 Chr.3: 46265227-46265227 on Build GRCh38
rs771039208 Chr.3: 46265131-46265133 on Build GRCh38
rs368253693 Chr.3: 46265193-46265193 on Build GRCh38
rs780722835 Chr.3: 46265148-46265148 on Build GRCh38
rs563391532 Chr.3: 46265071-46265071 on Build GRCh38
rs573526435 Chr.3: 46265232-46265232 on Build GRCh38
rs938683692 Chr.3: 46265046-46265046 on Build GRCh38
rs199797347 Chr.3: 46265215-46265215 on Build GRCh38
rs981852937 Chr.3: 46265181-46265181 on Build GRCh38
rs778898049 Chr.3: 46265207-46265208 on Build GRCh38
rs567921837 Chr.3: 46265062-46265062 on Build GRCh38
rs991484436 Chr.3: 46265081-46265081 on Build GRCh38
rs4987124 Chr.3: 46265151-46265151 on Build GRCh38
rs754155748 Chr.3: 46265115-46265115 on Build GRCh38
rs759339421 Chr.3: 46265107-46265107 on Build GRCh38
rs749388102 Chr.3: 46265118-46265118 on Build GRCh38
rs200517174 Chr.3: 46265202-46265202 on Build GRCh38
rs4987125 Chr.3: 46265220-46265220 on Build GRCh38
rs776687659 Chr.3: 46265212-46265212 on Build GRCh38
rs750564895 Chr.3: 46265140-46265140 on Build GRCh38
rs755441932 Chr.3: 46265165-46265165 on Build GRCh38
rs142492736 Chr.3: 46265196-46265196 on Build GRCh38
rs377752739 Chr.3: 46265110-46265110 on Build GRCh38
rs758927069 Chr.3: 46265141-46265141 on Build GRCh38
rs4987053 Chr.3: 46265209-46265209 on Build GRCh38
rs201063284 Chr.3: 46265184-46265184 on Build GRCh38
rs112916828 Chr.3: 46265105-46265105 on Build GRCh38
rs758559990 Chr.3: 46265221-46265221 on Build GRCh38
rs145868328 Chr.3: 46265216-46265216 on Build GRCh38
rs765398499 Chr.3: 46265130-46265130 on Build GRCh38
rs747879435 Chr.3: 46265189-46265189 on Build GRCh38
rs766991924 Chr.3: 46265224-46265224 on Build GRCh38
rs202034637 Chr.3: 46265190-46265190 on Build GRCh38
rs766331033 Chr.3: 46265104-46265104 on Build GRCh38
rs770795221 Chr.3: 46265201-46265201 on Build GRCh38
rs757262777 Chr.3: 46265125-46265125 on Build GRCh38
rs575612719 Chr.3: 46265112-46265112 on Build GRCh38

View Full Product Details