Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161222348-161222619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOA2
Entrez Gene ID
336
Gene Name
apolipoprotein A2
Gene Aliases
Apo-AII, ApoA-II, apoAII
UniGene
Hs.237658
Gene Chromosome Location
Chr.1: 161222293-161223628 on Build GRCh38
Transcripts
NM_001643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3997018 c.186-4C>T p.? Chr.1: 161222526-161222526 on Build GRCh38
7079301 c.208C>T p.Q70* Chr.1: 161222483-161222483 on Build GRCh38
6508071 c.203C>T p.S68L Chr.1: 161222505-161222505 on Build GRCh38
1491816 c.222C>A p.P74P Chr.1: 161222486-161222486 on Build GRCh38
898538 c.207G>T p.K69N Chr.1: 161222501-161222501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200191201 Chr.1: 161222541-161222541 on Build GRCh38
rs865834629 Chr.1: 161222414-161222415 on Build GRCh38
rs5088 Chr.1: 161222461-161222461 on Build GRCh38
rs760378915 Chr.1: 161222394-161222394 on Build GRCh38
rs770336880 Chr.1: 161222398-161222398 on Build GRCh38
rs6413454 Chr.1: 161222420-161222420 on Build GRCh38
rs765054986 Chr.1: 161222561-161222561 on Build GRCh38
rs774770928 Chr.1: 161222521-161222521 on Build GRCh38
rs752976925 Chr.1: 161222438-161222438 on Build GRCh38
rs1006991518 Chr.1: 161222399-161222399 on Build GRCh38
rs138954773 Chr.1: 161222437-161222437 on Build GRCh38
rs1029859927 Chr.1: 161222379-161222379 on Build GRCh38
rs937394706 Chr.1: 161222396-161222396 on Build GRCh38
rs138452270 Chr.1: 161222465-161222465 on Build GRCh38
rs767563719 Chr.1: 161222429-161222429 on Build GRCh38
rs779695910 Chr.1: 161222478-161222478 on Build GRCh38
rs538876594 Chr.1: 161222579-161222579 on Build GRCh38
rs775553389 Chr.1: 161222531-161222531 on Build GRCh38
rs773443521 Chr.1: 161222403-161222403 on Build GRCh38
rs200538010 Chr.1: 161222466-161222466 on Build GRCh38
rs12824 Chr.1: 161222467-161222467 on Build GRCh38
rs757032616 Chr.1: 161222456-161222456 on Build GRCh38
rs768425737 Chr.1: 161222492-161222492 on Build GRCh38
rs759575953 Chr.1: 161222422-161222422 on Build GRCh38
rs76162486 Chr.1: 161222523-161222523 on Build GRCh38
rs776287664 Chr.1: 161222498-161222498 on Build GRCh38
rs6413453 Chr.1: 161222526-161222526 on Build GRCh38
rs754055541 Chr.1: 161222548-161222548 on Build GRCh38
rs750382331 Chr.1: 161222566-161222566 on Build GRCh38
rs773788407 Chr.1: 161222569-161222578 on Build GRCh38
rs757039675 Chr.1: 161222560-161222560 on Build GRCh38
rs749636227 Chr.1: 161222513-161222513 on Build GRCh38
rs764978622 Chr.1: 161222462-161222462 on Build GRCh38
rs760691677 Chr.1: 161222538-161222538 on Build GRCh38
rs1028824039 Chr.1: 161222576-161222576 on Build GRCh38
rs1803091 Chr.1: 161222490-161222490 on Build GRCh38
rs36027516 Chr.1: 161222406-161222406 on Build GRCh38
rs778751126 Chr.1: 161222460-161222460 on Build GRCh38
rs371681039 Chr.1: 161222522-161222522 on Build GRCh38
rs369936134 Chr.1: 161222554-161222554 on Build GRCh38
rs763424301 Chr.1: 161222574-161222574 on Build GRCh38
rs756325696 Chr.1: 161222447-161222447 on Build GRCh38
rs775317995 Chr.1: 161222393-161222393 on Build GRCh38
rs940369794 Chr.1: 161222530-161222530 on Build GRCh38
rs753545559 Chr.1: 161222428-161222428 on Build GRCh38
rs745396210 Chr.1: 161222387-161222387 on Build GRCh38
rs763308512 Chr.1: 161222416-161222416 on Build GRCh38
rs890291989 Chr.1: 161222586-161222586 on Build GRCh38
rs771665149 Chr.1: 161222392-161222392 on Build GRCh38
rs766810017 Chr.1: 161222419-161222419 on Build GRCh38
rs777908160 Chr.1: 161222451-161222451 on Build GRCh38
rs771286488 Chr.1: 161222514-161222514 on Build GRCh38

View Full Product Details