Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14434140-14434401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF7IP
Entrez Gene ID
55729
Gene Name
activating transcription factor 7 interacting protein
Gene Aliases
AM, ATF-IP, MCAF, MCAF1, p621
UniGene
Hs.504856
Gene Chromosome Location
Chr.12: 14365632-14502935 on Build GRCh38
Transcripts
NM_001286514.1 NM_001286515.1 NM_018179.4 NM_181352.1 XM_005253424.3 XM_006719108.3 XM_006719109.3 XM_011520754.2 XM_011520755.2 XM_011520756.2 XM_017019638.1 XM_017019639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777968 c.1586A>G p.K529R Chr.12: 14434367-14434367 on Build GRCh38
937499 c.1593T>C p.P531P Chr.12: 14434371-14434371 on Build GRCh38
1128636 c.1589A>G p.K530R Chr.12: 14434367-14434367 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1012651152 Chr.12: 14434173-14434173 on Build GRCh38
rs775917225 Chr.12: 14434310-14434310 on Build GRCh38
rs556322838 Chr.12: 14434351-14434351 on Build GRCh38
rs151331582 Chr.12: 14434246-14434246 on Build GRCh38
rs1004303903 Chr.12: 14434203-14434203 on Build GRCh38
rs999644201 Chr.12: 14434267-14434267 on Build GRCh38
rs962968037 Chr.12: 14434228-14434228 on Build GRCh38
rs762280482 Chr.12: 14434333-14434333 on Build GRCh38
rs750216286 Chr.12: 14434186-14434186 on Build GRCh38
rs769937687 Chr.12: 14434290-14434290 on Build GRCh38
rs1028351607 Chr.12: 14434278-14434278 on Build GRCh38
rs774676266 Chr.12: 14434329-14434329 on Build GRCh38
rs764643907 Chr.12: 14434323-14434323 on Build GRCh38
rs372036691 Chr.12: 14434369-14434369 on Build GRCh38
rs902647822 Chr.12: 14434253-14434253 on Build GRCh38
rs1016262114 Chr.12: 14434227-14434227 on Build GRCh38
rs370379417 Chr.12: 14434355-14434355 on Build GRCh38
rs763454677 Chr.12: 14434316-14434316 on Build GRCh38
rs145246872 Chr.12: 14434364-14434364 on Build GRCh38
rs3213764 Chr.12: 14434367-14434367 on Build GRCh38
rs907420542 Chr.12: 14434180-14434180 on Build GRCh38
rs1023990717 Chr.12: 14434179-14434179 on Build GRCh38
rs974492440 Chr.12: 14434257-14434257 on Build GRCh38
rs933537700 Chr.12: 14434196-14434196 on Build GRCh38

View Full Product Details