Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62799329-62799556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGBP
Entrez Gene ID
55257
Gene Name
MRG/MORF4L binding protein
Gene Aliases
C20orf20, Eaf7, MRG15BP, URCC4
UniGene
Hs.443260
Gene Chromosome Location
Chr.20: 62796405-62801738 on Build GRCh38
Transcripts
NM_018270.5 NR_136405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554771 c.505G>C p.G169R Chr.20: 62799533-62799533 on Build GRCh38
6094205 c.525C>G p.D175E Chr.20: 62799553-62799553 on Build GRCh38
6845468 c.458C>T p.A153V Chr.20: 62799486-62799486 on Build GRCh38
6094206 c.470C>G p.S157C Chr.20: 62799498-62799498 on Build GRCh38
5486159 c.434C>T p.S145L Chr.20: 62799462-62799462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372719684 Chr.20: 62799385-62799386 on Build GRCh38
rs185858280 Chr.20: 62799522-62799522 on Build GRCh38
rs369232671 Chr.20: 62799412-62799412 on Build GRCh38
rs369295036 Chr.20: 62799406-62799406 on Build GRCh38
rs754740329 Chr.20: 62799408-62799408 on Build GRCh38
rs961288626 Chr.20: 62799403-62799403 on Build GRCh38
rs766064630 Chr.20: 62799529-62799529 on Build GRCh38
rs181588180 Chr.20: 62799441-62799441 on Build GRCh38
rs751195392 Chr.20: 62799535-62799535 on Build GRCh38
rs775851144 Chr.20: 62799481-62799481 on Build GRCh38
rs757311002 Chr.20: 62799430-62799430 on Build GRCh38
rs748071968 Chr.20: 62799428-62799428 on Build GRCh38
rs369540111 Chr.20: 62799504-62799504 on Build GRCh38
rs772837524 Chr.20: 62799515-62799515 on Build GRCh38
rs538269382 Chr.20: 62799364-62799364 on Build GRCh38
rs747417382 Chr.20: 62799487-62799487 on Build GRCh38
rs867434888 Chr.20: 62799375-62799375 on Build GRCh38
rs997345265 Chr.20: 62799361-62799361 on Build GRCh38
rs1015490160 Chr.20: 62799402-62799402 on Build GRCh38
rs554320509 Chr.20: 62799447-62799447 on Build GRCh38
rs919588218 Chr.20: 62799425-62799425 on Build GRCh38
rs764668275 Chr.20: 62799514-62799514 on Build GRCh38
rs746088074 Chr.20: 62799446-62799446 on Build GRCh38
rs533339951 Chr.20: 62799378-62799379 on Build GRCh38
rs76839923 Chr.20: 62799377-62799377 on Build GRCh38
rs137896518 Chr.20: 62799493-62799493 on Build GRCh38
rs762315674 Chr.20: 62799513-62799513 on Build GRCh38

View Full Product Details