Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76008487-76008718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
UniGene
Hs.500635
Gene Chromosome Location
Chr.17: 76006845-76027452 on Build GRCh38
Transcripts
NM_001320747.1 NM_001988.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5530656 c.4534G>A p.A1512T Chr.17: 76008671-76008671 on Build GRCh38
707171 c.4631A>G p.N1544S Chr.17: 76008574-76008574 on Build GRCh38
6656938 c.4495T>C p.C1499R Chr.17: 76008710-76008710 on Build GRCh38
6523905 c.4574G>T p.R1525L Chr.17: 76008631-76008631 on Build GRCh38
5530661 c.4619G>A p.W1540* Chr.17: 76008586-76008586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779417063 Chr.17: 76008589-76008589 on Build GRCh38
rs572601816 Chr.17: 76008615-76008615 on Build GRCh38
rs761341904 Chr.17: 76008530-76008530 on Build GRCh38
rs759305677 Chr.17: 76008632-76008632 on Build GRCh38
rs766845248 Chr.17: 76008686-76008686 on Build GRCh38
rs528132734 Chr.17: 76008688-76008688 on Build GRCh38
rs866995164 Chr.17: 76008652-76008652 on Build GRCh38
rs200073444 Chr.17: 76008671-76008671 on Build GRCh38
rs780341143 Chr.17: 76008593-76008593 on Build GRCh38
rs113394630 Chr.17: 76008519-76008519 on Build GRCh38
rs139365903 Chr.17: 76008647-76008647 on Build GRCh38
rs750335053 Chr.17: 76008657-76008659 on Build GRCh38
rs753898829 Chr.17: 76008509-76008509 on Build GRCh38
rs767807959 Chr.17: 76008504-76008504 on Build GRCh38
rs948686823 Chr.17: 76008537-76008537 on Build GRCh38
rs757772040 Chr.17: 76008510-76008510 on Build GRCh38
rs765264329 Chr.17: 76008646-76008647 on Build GRCh38
rs766131192 Chr.17: 76008547-76008547 on Build GRCh38
rs753917041 Chr.17: 76008573-76008573 on Build GRCh38
rs776295618 Chr.17: 76008529-76008529 on Build GRCh38
rs146378151 Chr.17: 76008523-76008523 on Build GRCh38
rs761906854 Chr.17: 76008643-76008643 on Build GRCh38
rs764329412 Chr.17: 76008643-76008643 on Build GRCh38
rs148554122 Chr.17: 76008532-76008532 on Build GRCh38
rs892895479 Chr.17: 76008602-76008602 on Build GRCh38
rs199755125 Chr.17: 76008557-76008557 on Build GRCh38
rs894998721 Chr.17: 76008580-76008580 on Build GRCh38
rs1052833673 Chr.17: 76008591-76008591 on Build GRCh38
rs755349309 Chr.17: 76008695-76008695 on Build GRCh38
rs150027671 Chr.17: 76008683-76008683 on Build GRCh38
rs751361738 Chr.17: 76008548-76008548 on Build GRCh38
rs867668690 Chr.17: 76008644-76008644 on Build GRCh38
rs147437042 Chr.17: 76008590-76008590 on Build GRCh38
rs769481718 Chr.17: 76008617-76008617 on Build GRCh38
rs201429207 Chr.17: 76008629-76008629 on Build GRCh38
rs774027925 Chr.17: 76008630-76008630 on Build GRCh38
rs377216983 Chr.17: 76008661-76008661 on Build GRCh38
rs144390976 Chr.17: 76008684-76008684 on Build GRCh38
rs768199812 Chr.17: 76008599-76008599 on Build GRCh38
rs757285861 Chr.17: 76008577-76008577 on Build GRCh38
rs564557598 Chr.17: 76008601-76008601 on Build GRCh38
rs534116655 Chr.17: 76008514-76008514 on Build GRCh38
rs751902650 Chr.17: 76008689-76008689 on Build GRCh38
rs776783322 Chr.17: 76008648-76008648 on Build GRCh38
rs897387869 Chr.17: 76008533-76008533 on Build GRCh38
rs754041039 Chr.17: 76008556-76008557 on Build GRCh38
rs746261242 Chr.17: 76008517-76008517 on Build GRCh38
rs144149788 Chr.17: 76008625-76008625 on Build GRCh38
rs747340763 Chr.17: 76008598-76008598 on Build GRCh38
rs867129386 Chr.17: 76008635-76008635 on Build GRCh38
rs966584955 Chr.17: 76008609-76008609 on Build GRCh38
rs200253222 Chr.17: 76008508-76008508 on Build GRCh38
rs944207499 Chr.17: 76008521-76008521 on Build GRCh38
rs535957560 Chr.17: 76008524-76008524 on Build GRCh38
rs1039147899 Chr.17: 76008643-76008645 on Build GRCh38
rs764241294 Chr.17: 76008567-76008567 on Build GRCh38
rs756324680 Chr.17: 76008505-76008505 on Build GRCh38
rs868094270 Chr.17: 76008631-76008631 on Build GRCh38
rs563372463 Chr.17: 76008536-76008536 on Build GRCh38

View Full Product Details