Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84565702-84565939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTBS
Entrez Gene ID
1486
Gene Name
chitobiase
Gene Aliases
CTB
UniGene
Hs.513557
Gene Chromosome Location
Chr.1: 84553121-84574480 on Build GRCh38
Transcripts
NM_004388.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4675297 c.613T>G p.F205V Chr.1: 84565925-84565925 on Build GRCh38
6448679 c.652G>C p.E218Q Chr.1: 84565886-84565886 on Build GRCh38
3751413 c.645C>A p.I215I Chr.1: 84565893-84565893 on Build GRCh38
5807743 c.697+3A>G p.? Chr.1: 84565838-84565838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760870585 Chr.1: 84565791-84565791 on Build GRCh38
rs373529980 Chr.1: 84565864-84565864 on Build GRCh38
rs905777605 Chr.1: 84565818-84565818 on Build GRCh38
rs762829389 Chr.1: 84565803-84565803 on Build GRCh38
rs748795511 Chr.1: 84565877-84565877 on Build GRCh38
rs917231438 Chr.1: 84565743-84565743 on Build GRCh38
rs779438848 Chr.1: 84565874-84565874 on Build GRCh38
rs545320856 Chr.1: 84565894-84565894 on Build GRCh38
rs557401356 Chr.1: 84565748-84565748 on Build GRCh38
rs781131584 Chr.1: 84565854-84565854 on Build GRCh38
rs895621046 Chr.1: 84565744-84565744 on Build GRCh38
rs926151224 Chr.1: 84565880-84565880 on Build GRCh38
rs536205798 Chr.1: 84565784-84565784 on Build GRCh38
rs771101356 Chr.1: 84565910-84565910 on Build GRCh38
rs750317091 Chr.1: 84565742-84565742 on Build GRCh38
rs41284599 Chr.1: 84565809-84565809 on Build GRCh38
rs764043260 Chr.1: 84565808-84565808 on Build GRCh38
rs748595949 Chr.1: 84565886-84565886 on Build GRCh38
rs755772693 Chr.1: 84565867-84565867 on Build GRCh38
rs773373898 Chr.1: 84565911-84565911 on Build GRCh38
rs80089235 Chr.1: 84565735-84565735 on Build GRCh38
rs757159462 Chr.1: 84565843-84565843 on Build GRCh38
rs766457857 Chr.1: 84565793-84565793 on Build GRCh38
rs1044345228 Chr.1: 84565811-84565811 on Build GRCh38
rs779510153 Chr.1: 84565882-84565882 on Build GRCh38
rs772480303 Chr.1: 84565890-84565890 on Build GRCh38
rs769070229 Chr.1: 84565879-84565879 on Build GRCh38
rs114528495 Chr.1: 84565893-84565893 on Build GRCh38
rs753887233 Chr.1: 84565800-84565800 on Build GRCh38
rs574053811 Chr.1: 84565752-84565752 on Build GRCh38

View Full Product Details